Gene Transcripts
- Gene Symbol
- ADAMTS9-AS2
- Entrez Gene ID
- 100507098
- Gene Name
- ADAMTS9 antisense RNA 2
- Gene Aliases
- Chromosome Location
- Chr. 3: 64684870 - 65011468 on Build GRCh38
- UniGene ID
- Hs.660881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_038264.1 |
|
2 |
694 |
|
GenBank mRNA |
AL832788.1 |
- |
2 |
655 |
|
BC040632.1 |
- |
2 |
436 |
|
View Full Product Details