Gene Transcripts
- Gene Symbol
- SLC25A51P1
- Entrez Gene ID
- 442229
- Gene Name
- solute carrier family 25 member 51 pseudogene 1
- Gene Aliases
- MCART3P, bA707M13.1
- Chromosome Location
- Chr. 6: 65787879 - 65789483 on Build GRCh38
- UniGene ID
- Hs.390414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026540.1 |
|
1 |
959 |
|
GenBank mRNA |
BC024198.1 |
- |
1 |
960 |
|
View Full Product Details