Gene Transcripts
- Gene Symbol
- ANP32A
- Entrez Gene ID
- 8125
- Gene Name
- acidic nuclear phosphoprotein 32 family member A
- Gene Aliases
- C15orf1, HPPCn, I1PP2A, LANP, MAPM, PHAP1, PHAPI, PP32
- Chromosome Location
- Chr. 15: 68778535 - 68820922 on Build GRCh38
- UniGene ID
- Hs.458747
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK127498.1 |
- |
1 |
360 |
|
View Full Product Details