Gene Transcripts
- Gene Symbol
- C6orf99
- Entrez Gene ID
- 100130967
- Gene Name
- chromosome 6 open reading frame 99
- Gene Aliases
- yR211F11.1
- Chromosome Location
- Chr. 6: 158869865 - 158918115 on Build GRCh38
- UniGene ID
- Hs.32804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195032.1 |
NP_001181961.1
|
2, 3 |
255 |
|
NM_001302839.1 |
NP_001289768.1
|
2, 3 |
589 |
|
NM_001302840.1 |
NP_001289769.1
|
3, 4 |
350 |
|
NM_001302841.1 |
NP_001289770.1
|
2, 3 |
249 |
|
XM_005266788.4 |
XP_005266845.1
|
2, 3 |
596 |
|
XM_017010142.1 |
XP_016865631.1
|
2, 3 |
186 |
|
XM_017010143.1 |
XP_016865632.1
|
2, 3 |
263 |
|
XM_017010144.1 |
XP_016865633.1
|
2, 3 |
258 |
|
XM_017010145.1 |
XP_016865634.1
|
1, 2 |
161 |
|
XM_017010146.1 |
XP_016865635.1
|
1, 2 |
156 |
|
XM_017010147.1 |
XP_016865636.1
|
2, 3 |
246 |
|
GenBank mRNA |
AA913820.1 |
- |
1, 2 |
396 |
|
AW138390.1 |
- |
1, 2 |
396 |
|
BF688381.1 |
- |
2, 3 |
256 |
|
HY205185.1 |
- |
1, 2 |
297 |
|
W37337.1 |
- |
2, 3 |
177 |
|
View Full Product Details