Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173793.4 | NP_776154.3 | 3 | 783 | |
GenBank mRNA | AK095271.1 | - | 3 | 243 | |
AK290073.1 | - | 3 | 756 | ||
BC062599.1 | - | 3 | 248 | 5248209 | |
BX337505.2 | - | 3 | 783 | ||
CR456339.1 | - | 3 | 244 |