Gene Transcripts
- Gene Symbol
- ANKRD29
- Entrez Gene ID
- 147463
- Gene Name
- ankyrin repeat domain 29
- Gene Aliases
- Chromosome Location
- Chr. 18: 23598926 - 23662914 on Build GRCh38
- UniGene ID
- Hs.355689
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001308238.1 |
NP_001295167.1
|
7, 8 |
786 |
|
NM_173505.3 |
NP_775776.2
|
7, 8 |
786 |
|
XM_011525830.2 |
XP_011524132.1
|
6, 7 |
706 |
|
XM_011525831.2 |
XP_011524133.1
|
6, 7 |
2726 |
|
XM_011525832.2 |
XP_011524134.1
|
6, 7 |
2844 |
|
XM_017025561.1 |
XP_016881050.1
|
6, 7 |
709 |
|
GenBank mRNA |
AK057782.1 |
- |
7, 8 |
716 |
|
AK123197.1 |
- |
4, 5 |
2106 |
|
AK313667.1 |
- |
7, 8 |
704 |
|
BC030622.1 |
- |
7, 8 |
791 |
4818619 |
View Full Product Details