Gene Transcripts

Gene Symbol
EIF4B
Entrez Gene ID
1975
Gene Name
eukaryotic translation initiation factor 4B
Gene Aliases
EIF-4B, PRO1843
Chromosome Location
Chr. 12: 53006258 - 53042209 on Build GRCh38
UniGene ID
Hs.648394
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300821.1 NP_001287750.1 7 968
NM_001417.5 NP_001408.2 7 968
GenBank mRNA AB076839.1 - 6 742
AK222528.1 - 7 770
AK299329.1 - 7 770
AK299526.1 - 6 654
BC073139.1 - 7 758 6527540
BC073154.1 - 7 758 6015183
CR456958.1 - 6 742

View Full Product Details