Gene Transcripts

Gene Symbol
ZBTB32
Entrez Gene ID
27033
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
FAXF, FAZF, Rog, TZFP, ZNF538
Chromosome Location
Chr. 19: 35704481 - 35717048 on Build GRCh38
UniGene ID
Hs.99430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316902.1 NP_001303831.1 4 336
NM_014383.2 NP_055198.1 4 1276
XM_011526718.2 XP_011525020.1 3 1437
XM_017026588.1 XP_016882077.1 2 942
XM_017026589.1 XP_016882078.1 4 1647
XM_017026591.1 XP_016882080.1 4 342
GenBank mRNA AF130255.1 - 3 1128
AF165097.1 - 2 918
HY039545.1 - 4 251

View Full Product Details