Gene Transcripts
- Gene Symbol
- C12orf42
- Entrez Gene ID
- 374470
- Gene Name
- chromosome 12 open reading frame 42
- Gene Aliases
- Chromosome Location
- Chr. 12: 103047669 - 103525258 on Build GRCh38
- UniGene ID
- Hs.534649
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001099336.2 |
NP_001092806.1
|
3, 4 |
413 |
|
NM_001278419.1 |
NP_001265348.1
|
3, 4 |
413 |
|
NM_001278420.1 |
NP_001265349.1
|
3, 4 |
278 |
|
NM_198521.3 |
NP_940923.2
|
3, 4 |
278 |
|
NR_103526.1 |
|
3, 4 |
278 |
|
XM_006719377.3 |
XP_006719440.1
|
2, 3 |
216 |
|
XM_011538291.1 |
XP_011536593.1
|
3, 4 |
273 |
|
XM_011538292.2 |
XP_011536594.1
|
3, 4 |
242 |
|
XM_011538293.2 |
XP_011536595.1
|
3, 4 |
249 |
|
XM_011538295.1 |
XP_011536597.1
|
3, 4 |
305 |
|
XM_011538306.1 |
XP_011536608.1
|
3, 4 |
273 |
|
XM_011538308.2 |
XP_011536610.1
|
4, 5 |
303 |
|
XM_011538312.2 |
XP_011536614.1
|
4, 5 |
301 |
|
XM_011538315.2 |
XP_011536617.1
|
4, 5 |
294 |
|
XM_017019276.1 |
XP_016874765.1
|
3, 4 |
367 |
|
XM_017019281.1 |
XP_016874770.1
|
4, 5 |
298 |
|
GenBank mRNA |
AK058052.1 |
- |
3, 4 |
236 |
|
BC039352.1 |
- |
3, 4 |
236 |
|
BC044617.1 |
- |
3, 4 |
374 |
5267756 |
BC117392.1 |
- |
3, 4 |
236 |
40125943 |
BC117394.1 |
- |
3, 4 |
236 |
40125945 |
BI830750.1 |
- |
3, 4 |
355 |
|
HY002043.1 |
- |
3, 4 |
371 |
|
HY015666.1 |
- |
3, 4 |
278 |
|
View Full Product Details