Gene Transcripts

Gene Symbol
MUC21
Entrez Gene ID
394263
Gene Name
mucin 21, cell surface associated
Gene Aliases
C6orf205, KMQK697, MUC-21
Chromosome Location
Chr. 6: 30983708 - 30988637 on Build GRCh38
UniGene ID
Hs.448277
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010909.3 NP_001010909.2 2 1629
NM_001322370.1 NP_001309299.1 1695
NM_001322371.1 NP_001309300.1 1802
NR_130720.1 2 1771
GenBank mRNA AB242595.1 - 2 1539
AK056612.1 - 1 124
AK298642.1 - 2 1617
AK299471.1 - 1802
AK301645.1 - 1 1337
AK316363.1 - 2 1619
AY358415.1 - 1706
BC105735.1 - 2 1564 40026480
BC105736.1 - 2 1706
BC105737.1 - 2 1564 40026476
BC107478.1 - 3 1114 40026469
HM596784.1 - 2 1558

View Full Product Details