Gene Transcripts

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
Chromosome Location
Chr. 12: 8102932 - 8138607 on Build GRCh38
UniGene ID
Hs.504657
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016184.3 NP_057268.1 4 644
NM_194447.2 NP_919429.2 3 527
NM_194448.2 NP_919430.1 2 428
NM_194450.2 NP_919432.1 3 545
XM_011520684.2 XP_011518986.1 3 1905
XM_017019382.1 XP_016874871.1 5 565
GenBank mRNA AF109146.1 - 4 491
AF200738.1 - 3 298
AF328684.1 - 4 636
AJ133532.1 - 4 638
BC074822.2 - 4 419 30915322
BC074874.2 - 4 419 30915470
BC117439.1 - 4 539 40125990
BC117441.1 - 4 539 40125992

View Full Product Details