Gene Transcripts

Gene Symbol
RPS6KB2
Entrez Gene ID
6199
Gene Name
ribosomal protein S6 kinase B2
Gene Aliases
KLS, P70-beta, P70-beta-1, P70-beta-2, S6K-beta2, S6K2, SRK, STK14B, p70(S6K)-beta, p70S6Kb
Chromosome Location
Chr. 11: 67428460 - 67435408 on Build GRCh38
UniGene ID
Hs.534345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003952.2 NP_003943.2 3 283
XM_005274164.1 XP_005274221.1 2 231
XM_005274165.4 XP_005274222.1 3 256
XM_006718655.3 XP_006718718.1 3 264
XM_006718656.3 XP_006718719.1 2 215
XM_006718657.1 XP_006718720.1 2 197
XM_017018108.1 XP_016873597.1 3 278
GenBank mRNA AB016869.1 - 3 255
AB019245.1 - 3 253
AB208827.1 - 1 697
AF076931.1 - 3 236
AF099739.1 - 3 240
AK297527.1 - 3 249
BC000094.2 - 3 256 3508140
BC136564.1 - 3 257 9020554
BX337895.2 - 3 286

View Full Product Details