Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022436.2 | NP_071881.1 | 5 | 691 | |
XM_005264480.3 | XP_005264537.1 | 5 | 712 | ||
XM_006712073.3 | XP_006712136.1 | 5 | 712 | ||
XM_006712074.3 | XP_006712137.1 | 5 | 712 | ||
XM_011533024.2 | XP_011531326.1 | 5 | 712 | ||
XM_011533025.2 | XP_011531327.1 | 5 | 1136 | ||
XM_011533026.2 | XP_011531328.1 | 5 | 712 | ||
XM_011533027.2 | XP_011531329.1 | 2 | 981 | ||
GenBank mRNA | AF312715.2 | - | 5 | 691 | |
AF320293.1 | - | 5 | 657 | ||
AK091997.1 | - | 4 | 1318 | ||
BC111541.1 | - | 4 | 1556 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_005264364.4 | XP_005264421.1 | 14 | 1894 | |
XM_005264365.4 | XP_005264422.1 | 14 | 1891 |