Gene Transcripts

Gene Symbol
ARIH2OS
Entrez Gene ID
646450
Gene Name
ariadne homolog 2 opposite strand
Gene Aliases
C3orf71
Chromosome Location
Chr. 3: 48917788 - 48919385 on Build GRCh38
UniGene ID
Hs.720727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123040.1 NP_001116512.1 1 939
GenBank mRNA AK097724.1 - 1 939
DA461567.1 - 1 213

View Full Product Details