Gene Transcripts

Gene Symbol
CAPNS1
Entrez Gene ID
826
Gene Name
calpain small subunit 1
Gene Aliases
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
Chromosome Location
Chr. 19: 36139926 - 36150353 on Build GRCh38
UniGene ID
Hs.515371
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003962.2 NP_001003962.1 8, 9 859
NM_001302632.1 NP_001289561.1 8, 9 766
NM_001302633.1 NP_001289562.1 7, 8 551
NM_001749.3 NP_001740.1 8, 9 862
XM_005259295.1 XP_005259352.1 8, 9 662
XM_005259296.1 XP_005259353.1 8, 9 665
XM_011527359.1 XP_011525661.1 7, 8 492
GenBank mRNA AK289606.1 - 8, 9 801
AY007141.1 - 8, 9 659
BC000592.2 - 8, 9 680 3347146
BC007779.2 - 8, 9 690 4301394
BC011903.2 - 8, 9 680 4120579
BC017308.2 - 8, 9 659 5089022
BC018931.2 - 8, 9 750 4303926
BC021933.1 - 8, 9 654 4872635
BC023643.2 - 8, 9 652 5017512
BC064998.1 - 8, 9 767 5785274
BF822052.1 - 2, 3 146
BQ891655.1 - 5, 6 323
BT009775.1 - 7, 8 615
CN349928.1 - 7, 8 500
X04106.1 - 8, 9 773

View Full Product Details