Gene Transcripts

Gene Symbol
DYSF
Entrez Gene ID
8291
Gene Name
dysferlin
Gene Aliases
FER1L1, LGMD2B, MMD1
Chromosome Location
Chr. 2: 71453155 - 71686763 on Build GRCh38
UniGene ID
Hs.252180
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130455.1 NP_001123927.1 14, 15 1486
NM_001130976.1 NP_001124448.1 14, 15 1718
NM_001130977.1 NP_001124449.1 14, 15 1718
NM_001130978.1 NP_001124450.1 14, 15 1718
NM_001130979.1 NP_001124451.1 15, 16 1811
NM_001130980.1 NP_001124452.1 15, 16 1811
NM_001130981.1 NP_001124453.1 15, 16 1811
NM_001130982.1 NP_001124454.1 15, 16 1579
NM_001130983.1 NP_001124455.1 14, 15 1486
NM_001130984.1 NP_001124456.1 14, 15 1486
NM_001130985.1 NP_001124457.1 15, 16 1579
NM_001130986.1 NP_001124458.1 14, 15 1486
NM_001130987.1 NP_001124459.1 15, 16 1579
NM_003494.3 NP_003485.1 14, 15 1718
XM_005264584.4 XP_005264641.1 15, 16 1594
XM_005264585.4 XP_005264642.1 15, 16 2279
GenBank mRNA AF075575.1 - 14, 15 1715
AK074104.1 - 1, 2 30
BC171803.1 - 14, 15 1691
DQ267935.1 - 14, 15 1486
EU515155.1 - 15, 16 1712
EU515156.1 - 14, 15 1619
EU515157.1 - 14, 15 1619
EU515158.1 - 15, 16 1712
EU515159.1 - 14, 15 1619
EU515160.1 - 15, 16 1712
EU515161.1 - 15, 16 1579
EU515162.1 - 14, 15 1486
EU515163.1 - 14, 15 1486
EU515164.1 - 15, 16 1579
EU515165.1 - 14, 15 1486
EU515166.1 - 15, 16 1579

View Full Product Details