Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75003197-75003696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477773 c.331A>T p.K111* Chr.14: 75003597-75003597 on Build GRCh38
433375 c.301G>A p.D101N Chr.14: 75003567-75003567 on Build GRCh38
7313744 c.420G>T p.L140L Chr.14: 75003686-75003686 on Build GRCh38
7121593 c.353T>G p.L118R Chr.14: 75003619-75003619 on Build GRCh38
5075305 c.195C>G p.G65G Chr.14: 75003306-75003306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777252700 Chr.14: 75003500-75003500 on Build GRCh38
rs907086619 Chr.14: 75003308-75003308 on Build GRCh38
rs1056310003 Chr.14: 75003393-75003393 on Build GRCh38
rs762604029 Chr.14: 75003543-75003543 on Build GRCh38
rs377750951 Chr.14: 75003244-75003244 on Build GRCh38
rs377659896 Chr.14: 75003353-75003353 on Build GRCh38
rs372208258 Chr.14: 75003246-75003246 on Build GRCh38
rs372174903 Chr.14: 75003650-75003650 on Build GRCh38
rs368133564 Chr.14: 75003564-75003564 on Build GRCh38
rs761372299 Chr.14: 75003249-75003249 on Build GRCh38
rs758611010 Chr.14: 75003293-75003293 on Build GRCh38
rs767584462 Chr.14: 75003418-75003418 on Build GRCh38
rs766278140 Chr.14: 75003255-75003255 on Build GRCh38
rs759621968 Chr.14: 75003599-75003599 on Build GRCh38
rs142827960 Chr.14: 75003345-75003345 on Build GRCh38
rs752913982 Chr.14: 75003283-75003283 on Build GRCh38
rs751634475 Chr.14: 75003297-75003297 on Build GRCh38
rs746678040 Chr.14: 75003512-75003512 on Build GRCh38
rs1039929308 Chr.14: 75003643-75003643 on Build GRCh38
rs775335483 Chr.14: 75003537-75003537 on Build GRCh38
rs374195020 Chr.14: 75003620-75003620 on Build GRCh38
rs764275551 Chr.14: 75003399-75003399 on Build GRCh38
rs759366253 Chr.14: 75003262-75003262 on Build GRCh38
rs772745651 Chr.14: 75003343-75003343 on Build GRCh38
rs777077732 Chr.14: 75003666-75003668 on Build GRCh38
rs397514648 Chr.14: 75003365-75003365 on Build GRCh38
rs776342453 Chr.14: 75003522-75003522 on Build GRCh38
rs769894290 Chr.14: 75003532-75003532 on Build GRCh38
rs183116394 Chr.14: 75003437-75003437 on Build GRCh38
rs373906805 Chr.14: 75003550-75003550 on Build GRCh38
rs1020267453 Chr.14: 75003468-75003468 on Build GRCh38
rs768864027 Chr.14: 75003521-75003522 on Build GRCh38
rs779349624 Chr.14: 75003611-75003611 on Build GRCh38
rs768524274 Chr.14: 75003544-75003544 on Build GRCh38
rs751159755 Chr.14: 75003612-75003612 on Build GRCh38
rs200073232 Chr.14: 75003411-75003411 on Build GRCh38
rs200642949 Chr.14: 75003648-75003648 on Build GRCh38
rs201912801 Chr.14: 75003671-75003671 on Build GRCh38
rs746456163 Chr.14: 75003350-75003350 on Build GRCh38
rs755232669 Chr.14: 75003608-75003608 on Build GRCh38
rs747822193 Chr.14: 75003337-75003337 on Build GRCh38
rs757757428 Chr.14: 75003307-75003307 on Build GRCh38
rs373932168 Chr.14: 75003528-75003528 on Build GRCh38
rs745423619 Chr.14: 75003633-75003633 on Build GRCh38
rs780391000 Chr.14: 75003434-75003434 on Build GRCh38
rs772654412 Chr.14: 75003254-75003254 on Build GRCh38
rs886050765 Chr.14: 75003382-75003382 on Build GRCh38
rs771410713 Chr.14: 75003227-75003227 on Build GRCh38
rs775188722 Chr.14: 75003349-75003349 on Build GRCh38
rs770508411 Chr.14: 75003518-75003518 on Build GRCh38
rs531866161 Chr.14: 75003252-75003252 on Build GRCh38
rs756670980 Chr.14: 75003429-75003429 on Build GRCh38
rs967630584 Chr.14: 75003491-75003491 on Build GRCh38
rs764232354 Chr.14: 75003294-75003294 on Build GRCh38
rs370483997 Chr.14: 75003591-75003591 on Build GRCh38
rs755393155 Chr.14: 75003440-75003440 on Build GRCh38
rs765129237 Chr.14: 75003263-75003263 on Build GRCh38
rs150617429 Chr.14: 75003646-75003646 on Build GRCh38
rs771686725 Chr.14: 75003342-75003342 on Build GRCh38
rs918318919 Chr.14: 75003464-75003464 on Build GRCh38
rs17849324 Chr.14: 75003565-75003565 on Build GRCh38
rs1030584724 Chr.14: 75003506-75003506 on Build GRCh38
rs997727777 Chr.14: 75003501-75003501 on Build GRCh38
rs369511014 Chr.14: 75003324-75003324 on Build GRCh38
rs762017222 Chr.14: 75003673-75003673 on Build GRCh38
rs201622896 Chr.14: 75003667-75003667 on Build GRCh38
rs763206533 Chr.14: 75003398-75003398 on Build GRCh38
rs769455036 Chr.14: 75003638-75003638 on Build GRCh38
rs756401063 Chr.14: 75003331-75003331 on Build GRCh38
rs1038016772 Chr.14: 75003229-75003229 on Build GRCh38
rs773949162 Chr.14: 75003247-75003247 on Build GRCh38
rs781626544 Chr.14: 75003317-75003317 on Build GRCh38
rs368343040 Chr.14: 75003621-75003621 on Build GRCh38
rs770598243 Chr.14: 75003352-75003352 on Build GRCh38
rs750868104 Chr.14: 75003424-75003424 on Build GRCh38
rs751826162 Chr.14: 75003407-75003407 on Build GRCh38
rs929643497 Chr.14: 75003669-75003669 on Build GRCh38
rs766843755 Chr.14: 75003577-75003577 on Build GRCh38
rs538163666 Chr.14: 75003372-75003372 on Build GRCh38
rs750568077 Chr.14: 75003557-75003557 on Build GRCh38
rs200364116 Chr.14: 75003397-75003397 on Build GRCh38
rs762068601 Chr.14: 75003547-75003547 on Build GRCh38
rs141814339 Chr.14: 75003259-75003259 on Build GRCh38
rs780409528 Chr.14: 75003333-75003333 on Build GRCh38
rs112768305 Chr.14: 75003236-75003236 on Build GRCh38
rs376049144 Chr.14: 75003549-75003549 on Build GRCh38

View Full Product Details