Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75004624-75005059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362728 c.541G>A p.A181T Chr.14: 75004844-75004844 on Build GRCh38
5382676 c.515G>A p.R172Q Chr.14: 75004818-75004818 on Build GRCh38
699380 c.534C>G p.L178L Chr.14: 75004837-75004837 on Build GRCh38
5054103 c.544G>A p.A182T Chr.14: 75004847-75004847 on Build GRCh38
7107133 c.441A>C p.T147T Chr.14: 75004744-75004744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756483469 Chr.14: 75004943-75004945 on Build GRCh38
rs996519996 Chr.14: 75004972-75004972 on Build GRCh38
rs778773167 Chr.14: 75004720-75004720 on Build GRCh38
rs756957023 Chr.14: 75004966-75004966 on Build GRCh38
rs751963903 Chr.14: 75004692-75004693 on Build GRCh38
rs776279598 Chr.14: 75004903-75004903 on Build GRCh38
rs771150389 Chr.14: 75004690-75004706 on Build GRCh38
rs754470588 Chr.14: 75004788-75004788 on Build GRCh38
rs201498750 Chr.14: 75004691-75004691 on Build GRCh38
rs764725120 Chr.14: 75004920-75004920 on Build GRCh38
rs768704231 Chr.14: 75004695-75004695 on Build GRCh38
rs35602124 Chr.14: 75004750-75004750 on Build GRCh38
rs371500682 Chr.14: 75004700-75004700 on Build GRCh38
rs780858342 Chr.14: 75004946-75004946 on Build GRCh38
rs763973123 Chr.14: 75004938-75004938 on Build GRCh38
rs753454455 Chr.14: 75004787-75004787 on Build GRCh38
rs201252172 Chr.14: 75004702-75004702 on Build GRCh38
rs771082903 Chr.14: 75004719-75004727 on Build GRCh38
rs943408810 Chr.14: 75005032-75005032 on Build GRCh38
rs769799120 Chr.14: 75004731-75004731 on Build GRCh38
rs11628782 Chr.14: 75004794-75004794 on Build GRCh38
rs753426356 Chr.14: 75004716-75004716 on Build GRCh38
rs759931855 Chr.14: 75004701-75004701 on Build GRCh38
rs71426992 Chr.14: 75004705-75004705 on Build GRCh38
rs376012405 Chr.14: 75004690-75004696 on Build GRCh38
rs751422883 Chr.14: 75004831-75004831 on Build GRCh38
rs779627622 Chr.14: 75004805-75004805 on Build GRCh38
rs757027574 Chr.14: 75004839-75004839 on Build GRCh38
rs770130444 Chr.14: 75004705-75004706 on Build GRCh38
rs764926377 Chr.14: 75004690-75004691 on Build GRCh38
rs763372886 Chr.14: 75004704-75004704 on Build GRCh38
rs776167952 Chr.14: 75004703-75004703 on Build GRCh38
rs369078986 Chr.14: 75004851-75004851 on Build GRCh38
rs748617486 Chr.14: 75004704-75004705 on Build GRCh38
rs547439570 Chr.14: 75004886-75004886 on Build GRCh38
rs199617675 Chr.14: 75004693-75004693 on Build GRCh38
rs547821892 Chr.14: 75004712-75004718 on Build GRCh38
rs778540848 Chr.14: 75004696-75004696 on Build GRCh38
rs763187125 Chr.14: 75004688-75004692 on Build GRCh38
rs104894427 Chr.14: 75004850-75004850 on Build GRCh38
rs763771776 Chr.14: 75004690-75004703 on Build GRCh38
rs758398310 Chr.14: 75004817-75004817 on Build GRCh38
rs774920663 Chr.14: 75004755-75004755 on Build GRCh38
rs968595472 Chr.14: 75004876-75004876 on Build GRCh38
rs777605073 Chr.14: 75004725-75004725 on Build GRCh38
rs28592148 Chr.14: 75004692-75004692 on Build GRCh38
rs570430118 Chr.14: 75004953-75004953 on Build GRCh38
rs899404567 Chr.14: 75004669-75004669 on Build GRCh38
rs761363779 Chr.14: 75004690-75004701 on Build GRCh38
rs747246029 Chr.14: 75004699-75004699 on Build GRCh38
rs1046967315 Chr.14: 75004847-75004847 on Build GRCh38
rs910615977 Chr.14: 75004957-75004957 on Build GRCh38
rs765855445 Chr.14: 75004796-75004797 on Build GRCh38
rs760213491 Chr.14: 75004758-75004758 on Build GRCh38
rs759168949 Chr.14: 75004910-75004910 on Build GRCh38
rs747986323 Chr.14: 75004697-75004697 on Build GRCh38
rs774832957 Chr.14: 75004885-75004885 on Build GRCh38
rs753161850 Chr.14: 75004937-75004937 on Build GRCh38
rs750033749 Chr.14: 75004707-75004707 on Build GRCh38
rs867014078 Chr.14: 75004687-75004687 on Build GRCh38
rs200995477 Chr.14: 75004726-75004726 on Build GRCh38
rs745915599 Chr.14: 75004730-75004730 on Build GRCh38
rs868421455 Chr.14: 75004685-75004685 on Build GRCh38
rs139680549 Chr.14: 75004801-75004801 on Build GRCh38
rs752245327 Chr.14: 75004927-75004927 on Build GRCh38
rs775503992 Chr.14: 75004690-75004705 on Build GRCh38
rs552202441 Chr.14: 75004656-75004656 on Build GRCh38
rs746086054 Chr.14: 75004844-75004844 on Build GRCh38
rs113994011 Chr.14: 75004889-75004889 on Build GRCh38
rs760495653 Chr.14: 75004719-75004726 on Build GRCh38
rs760680969 Chr.14: 75004690-75004702 on Build GRCh38
rs777514938 Chr.14: 75004824-75004824 on Build GRCh38
rs776069996 Chr.14: 75004733-75004733 on Build GRCh38
rs377720530 Chr.14: 75004942-75004942 on Build GRCh38
rs796921417 Chr.14: 75004689-75004689 on Build GRCh38
rs781754516 Chr.14: 75004901-75004902 on Build GRCh38
rs113994010 Chr.14: 75004851-75004851 on Build GRCh38
rs763563473 Chr.14: 75004734-75004734 on Build GRCh38
rs993643728 Chr.14: 75004745-75004745 on Build GRCh38
rs1032427451 Chr.14: 75004975-75004975 on Build GRCh38
rs899125257 Chr.14: 75004818-75004818 on Build GRCh38
rs200354660 Chr.14: 75004698-75004698 on Build GRCh38
rs762858558 Chr.14: 75004935-75004935 on Build GRCh38
rs762617704 Chr.14: 75004730-75004730 on Build GRCh38
rs750034821 Chr.14: 75004725-75004725 on Build GRCh38
rs766102981 Chr.14: 75004765-75004765 on Build GRCh38
rs954527096 Chr.14: 75004904-75004904 on Build GRCh38
rs977260592 Chr.14: 75004880-75004880 on Build GRCh38
rs749527878 Chr.14: 75004707-75004719 on Build GRCh38
rs749806505 Chr.14: 75004873-75004873 on Build GRCh38
rs755436800 Chr.14: 75004872-75004872 on Build GRCh38
rs1037987278 Chr.14: 75004664-75004664 on Build GRCh38
rs770181233 Chr.14: 75004687-75004688 on Build GRCh38
rs757014213 Chr.14: 75004944-75004944 on Build GRCh38
rs1047855420 Chr.14: 75004753-75004753 on Build GRCh38
rs774535685 Chr.14: 75004719-75004720 on Build GRCh38
rs778316802 Chr.14: 75004950-75004950 on Build GRCh38
rs182322399 Chr.14: 75004721-75004721 on Build GRCh38
rs754847169 Chr.14: 75004719-75004719 on Build GRCh38
rs764538257 Chr.14: 75004744-75004744 on Build GRCh38
rs376803283 Chr.14: 75004727-75004727 on Build GRCh38
rs104894428 Chr.14: 75004815-75004815 on Build GRCh38
rs765799823 Chr.14: 75004708-75004708 on Build GRCh38
rs768701906 Chr.14: 75004720-75004721 on Build GRCh38
rs749163540 Chr.14: 75004694-75004694 on Build GRCh38
rs963775899 Chr.14: 75004956-75004962 on Build GRCh38
rs781440489 Chr.14: 75004843-75004843 on Build GRCh38

View Full Product Details