Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75009193-75009702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

SNP Detail

SNP ID Chromosome Location
rs893731760 Chr.14: 75009499-75009499 on Build GRCh38
rs984607853 Chr.14: 75009239-75009239 on Build GRCh38
rs1016850358 Chr.14: 75009653-75009653 on Build GRCh38
rs1046595426 Chr.14: 75009429-75009429 on Build GRCh38
rs1053417390 Chr.14: 75009472-75009472 on Build GRCh38
rs1017415169 Chr.14: 75009676-75009676 on Build GRCh38
rs760167860 Chr.14: 75009228-75009228 on Build GRCh38
rs954066524 Chr.14: 75009285-75009285 on Build GRCh38
rs998007996 Chr.14: 75009586-75009586 on Build GRCh38
rs1028392498 Chr.14: 75009277-75009277 on Build GRCh38
rs896076459 Chr.14: 75009548-75009548 on Build GRCh38
rs199523469 Chr.14: 75009236-75009236 on Build GRCh38
rs569575124 Chr.14: 75009448-75009448 on Build GRCh38
rs555158437 Chr.14: 75009602-75009602 on Build GRCh38
rs950237622 Chr.14: 75009417-75009417 on Build GRCh38
rs760892259 Chr.14: 75009325-75009328 on Build GRCh38
rs141381011 Chr.14: 75009576-75009576 on Build GRCh38
rs116393177 Chr.14: 75009244-75009244 on Build GRCh38
rs878922788 Chr.14: 75009302-75009302 on Build GRCh38
rs904219710 Chr.14: 75009424-75009424 on Build GRCh38
rs904920315 Chr.14: 75009449-75009449 on Build GRCh38
rs971977552 Chr.14: 75009256-75009256 on Build GRCh38
rs568593225 Chr.14: 75009603-75009603 on Build GRCh38
rs541027518 Chr.14: 75009430-75009431 on Build GRCh38
rs375056192 Chr.14: 75009221-75009221 on Build GRCh38
rs866429489 Chr.14: 75009649-75009649 on Build GRCh38
rs560892827 Chr.14: 75009254-75009254 on Build GRCh38
rs746045033 Chr.14: 75009475-75009475 on Build GRCh38
rs917444452 Chr.14: 75009407-75009407 on Build GRCh38
rs542050883 Chr.14: 75009362-75009362 on Build GRCh38
rs763277044 Chr.14: 75009414-75009416 on Build GRCh38
rs4556 Chr.14: 75009368-75009368 on Build GRCh38
rs187228528 Chr.14: 75009347-75009347 on Build GRCh38
rs3208685 Chr.14: 75009298-75009298 on Build GRCh38
rs1051612401 Chr.14: 75009343-75009343 on Build GRCh38
rs758861979 Chr.14: 75009599-75009599 on Build GRCh38
rs566966733 Chr.14: 75009357-75009357 on Build GRCh38
rs765822635 Chr.14: 75009233-75009233 on Build GRCh38
rs1058367 Chr.14: 75009556-75009556 on Build GRCh38
rs901033133 Chr.14: 75009581-75009581 on Build GRCh38
rs530228507 Chr.14: 75009346-75009346 on Build GRCh38
rs750077642 Chr.14: 75009226-75009226 on Build GRCh38
rs937674386 Chr.14: 75009465-75009465 on Build GRCh38
rs1009940184 Chr.14: 75009554-75009554 on Build GRCh38
rs372474383 Chr.14: 75009226-75009227 on Build GRCh38
rs766944047 Chr.14: 75009237-75009239 on Build GRCh38
rs984061062 Chr.14: 75009327-75009327 on Build GRCh38
rs552563342 Chr.14: 75009369-75009369 on Build GRCh38
rs970621931 Chr.14: 75009652-75009652 on Build GRCh38
rs3180290 Chr.14: 75009589-75009589 on Build GRCh38
rs139949391 Chr.14: 75009657-75009657 on Build GRCh38

View Full Product Details