Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1363247-1363776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNPTG
Entrez Gene ID
84572
Gene Name
N-acetylglucosamine-1-phosphate transferase gamma subunit
Gene Aliases
C16orf27, GNPTAG, LP2537, RJD9
UniGene
Hs.241575
Gene Chromosome Location
Chr.16: 1351899-1363351 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1 NM_001193388.3 NM_001193389.1 NM_001276414.1 XM_005255505.1 XM_011522610.1 XM_011522611.1 XM_011522612.1 XM_017023568.1 XM_017023569.1

Gene Symbol
UNKL
Entrez Gene ID
64718
Gene Name
unkempt family like zinc finger
Gene Aliases
C16orf28, ZC3H5L, ZC3HDC5L
UniGene
Hs.643536
Gene Chromosome Location
Chr.16: 1363205-1414749 on Build GRCh38
Transcripts
NM_032520.4 XM_017023782.1 XM_017023783.1 NM_001193388.3 NM_001193389.1 NM_001276414.1 XM_005255505.1 XM_011522610.1 XM_011522611.1 XM_011522612.1 XM_017023568.1 XM_017023569.1

SNP Detail

SNP ID Chromosome Location
rs1018283678 Chr.16: 1363352-1363352 on Build GRCh38
rs544096810 Chr.16: 1363423-1363423 on Build GRCh38
rs938807912 Chr.16: 1363468-1363468 on Build GRCh38
rs929903930 Chr.16: 1363630-1363630 on Build GRCh38
rs994097617 Chr.16: 1363296-1363296 on Build GRCh38
rs899713613 Chr.16: 1363535-1363535 on Build GRCh38
rs928904342 Chr.16: 1363462-1363462 on Build GRCh38
rs898155441 Chr.16: 1363502-1363502 on Build GRCh38
rs1033815943 Chr.16: 1363580-1363580 on Build GRCh38
rs771445582 Chr.16: 1363623-1363623 on Build GRCh38
rs547537104 Chr.16: 1363560-1363560 on Build GRCh38
rs964103294 Chr.16: 1363379-1363379 on Build GRCh38
rs1038326897 Chr.16: 1363541-1363541 on Build GRCh38
rs758351508 Chr.16: 1363380-1363380 on Build GRCh38
rs967816617 Chr.16: 1363445-1363445 on Build GRCh38
rs549149015 Chr.16: 1363643-1363643 on Build GRCh38
rs796860804 Chr.16: 1363591-1363591 on Build GRCh38
rs757707991 Chr.16: 1363282-1363282 on Build GRCh38
rs886051686 Chr.16: 1363296-1363296 on Build GRCh38
rs774503075 Chr.16: 1363474-1363475 on Build GRCh38
rs1016643337 Chr.16: 1363441-1363441 on Build GRCh38
rs550237556 Chr.16: 1363564-1363564 on Build GRCh38
rs532973425 Chr.16: 1363592-1363592 on Build GRCh38
rs1008041077 Chr.16: 1363325-1363325 on Build GRCh38
rs575854401 Chr.16: 1363287-1363287 on Build GRCh38
rs919694735 Chr.16: 1363642-1363642 on Build GRCh38
rs1003633771 Chr.16: 1363574-1363574 on Build GRCh38
rs943953468 Chr.16: 1363676-1363676 on Build GRCh38
rs56026628 Chr.16: 1363695-1363695 on Build GRCh38
rs991670410 Chr.16: 1363474-1363474 on Build GRCh38
rs535518208 Chr.16: 1363698-1363698 on Build GRCh38
rs868068574 Chr.16: 1363537-1363537 on Build GRCh38
rs897039334 Chr.16: 1363627-1363627 on Build GRCh38
rs769652235 Chr.16: 1363498-1363498 on Build GRCh38
rs886051687 Chr.16: 1363348-1363349 on Build GRCh38
rs963443026 Chr.16: 1363442-1363442 on Build GRCh38
rs1022898558 Chr.16: 1363690-1363690 on Build GRCh38
rs779865354 Chr.16: 1363505-1363505 on Build GRCh38
rs938259713 Chr.16: 1363292-1363292 on Build GRCh38
rs914916934 Chr.16: 1363305-1363305 on Build GRCh38
rs372220568 Chr.16: 1363638-1363638 on Build GRCh38
rs895299390 Chr.16: 1363662-1363662 on Build GRCh38
rs1007980455 Chr.16: 1363515-1363515 on Build GRCh38
rs998833896 Chr.16: 1363700-1363700 on Build GRCh38
rs538371215 Chr.16: 1363679-1363679 on Build GRCh38
rs539354203 Chr.16: 1363347-1363348 on Build GRCh38
rs113716484 Chr.16: 1363694-1363694 on Build GRCh38
rs1011847773 Chr.16: 1363691-1363691 on Build GRCh38
rs977689038 Chr.16: 1363452-1363452 on Build GRCh38
rs955080003 Chr.16: 1363757-1363757 on Build GRCh38
rs970870346 Chr.16: 1363453-1363453 on Build GRCh38
rs541466628 Chr.16: 1363506-1363506 on Build GRCh38
rs977761329 Chr.16: 1363603-1363604 on Build GRCh38
rs540794095 Chr.16: 1363374-1363374 on Build GRCh38
rs555685957 Chr.16: 1363278-1363278 on Build GRCh38
rs891062257 Chr.16: 1363508-1363508 on Build GRCh38
rs574237114 Chr.16: 1363484-1363484 on Build GRCh38
rs569695686 Chr.16: 1363585-1363585 on Build GRCh38
rs866134927 Chr.16: 1363302-1363302 on Build GRCh38
rs7192408 Chr.16: 1363315-1363315 on Build GRCh38
rs546732886 Chr.16: 1363605-1363605 on Build GRCh38
rs566657373 Chr.16: 1363607-1363607 on Build GRCh38
rs567324115 Chr.16: 1363505-1363506 on Build GRCh38
rs747157191 Chr.16: 1363488-1363488 on Build GRCh38
rs985209832 Chr.16: 1363620-1363620 on Build GRCh38
rs770118467 Chr.16: 1363487-1363487 on Build GRCh38
rs144083031 Chr.16: 1363351-1363351 on Build GRCh38
rs1029275494 Chr.16: 1363712-1363712 on Build GRCh38
rs899142213 Chr.16: 1363551-1363551 on Build GRCh38
rs761320112 Chr.16: 1363608-1363608 on Build GRCh38
rs993538512 Chr.16: 1363554-1363554 on Build GRCh38
rs1026420715 Chr.16: 1363446-1363446 on Build GRCh38
rs1005417140 Chr.16: 1363525-1363525 on Build GRCh38
rs561345340 Chr.16: 1363556-1363556 on Build GRCh38
rs963917300 Chr.16: 1363621-1363621 on Build GRCh38
rs569260717 Chr.16: 1363644-1363644 on Build GRCh38
rs890946572 Chr.16: 1363324-1363324 on Build GRCh38
rs535736709 Chr.16: 1363622-1363622 on Build GRCh38
rs912438255 Chr.16: 1363666-1363666 on Build GRCh38
rs889623314 Chr.16: 1363516-1363516 on Build GRCh38
rs1039513704 Chr.16: 1363534-1363534 on Build GRCh38
rs898367744 Chr.16: 1363284-1363284 on Build GRCh38
rs970780381 Chr.16: 1363565-1363565 on Build GRCh38
rs899662904 Chr.16: 1363682-1363682 on Build GRCh38
rs116345826 Chr.16: 1363426-1363426 on Build GRCh38
rs117222630 Chr.16: 1363749-1363749 on Build GRCh38
rs889675750 Chr.16: 1363392-1363392 on Build GRCh38
rs915518880 Chr.16: 1363586-1363586 on Build GRCh38
rs1038677405 Chr.16: 1363626-1363626 on Build GRCh38
rs867097744 Chr.16: 1363454-1363454 on Build GRCh38
rs796293830 Chr.16: 1363370-1363370 on Build GRCh38
rs181148746 Chr.16: 1363312-1363312 on Build GRCh38

View Full Product Details