Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40301133-40301627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL9A2
Entrez Gene ID
1298
Gene Name
collagen type IX alpha 2
Gene Aliases
DJ39G22.4, EDM2, MED, STL5
UniGene
Hs.418012
Gene Chromosome Location
Chr.1: 40300487-40317653 on Build GRCh38
Transcripts
NM_001852.3 XM_006710365.3 XM_011540715.2 XM_011540716.2 XM_011540717.2 XM_017000332.1 XM_017000333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3489540 c.2020G>A p.A674T Chr.1: 40301232-40301232 on Build GRCh38
909430 c.1952G>A p.R651Q Chr.1: 40301300-40301300 on Build GRCh38
3733637 c.1944_1945insAG p.A649fs*>42 Chr.1: 40301307-40301308 on Build GRCh38
1177136 c.1918C>T p.R640* Chr.1: 40301334-40301334 on Build GRCh38
3719932 c.1921G>C p.G641R Chr.1: 40301331-40301331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779177140 Chr.1: 40301309-40301309 on Build GRCh38
rs754673796 Chr.1: 40301291-40301291 on Build GRCh38
rs768840439 Chr.1: 40301349-40301349 on Build GRCh38
rs921184351 Chr.1: 40301307-40301307 on Build GRCh38
rs748710645 Chr.1: 40301422-40301426 on Build GRCh38
rs765856534 Chr.1: 40301284-40301284 on Build GRCh38
rs968288876 Chr.1: 40301228-40301228 on Build GRCh38
rs2229826 Chr.1: 40301251-40301251 on Build GRCh38
rs939837045 Chr.1: 40301490-40301490 on Build GRCh38
rs113002355 Chr.1: 40301542-40301543 on Build GRCh38
rs775823584 Chr.1: 40301176-40301176 on Build GRCh38
rs749873890 Chr.1: 40301202-40301202 on Build GRCh38
rs765273618 Chr.1: 40301201-40301201 on Build GRCh38
rs200971976 Chr.1: 40301370-40301370 on Build GRCh38
rs192569174 Chr.1: 40301544-40301544 on Build GRCh38
rs148008235 Chr.1: 40301233-40301233 on Build GRCh38
rs1001015047 Chr.1: 40301552-40301552 on Build GRCh38
rs904030663 Chr.1: 40301539-40301539 on Build GRCh38
rs746019124 Chr.1: 40301163-40301163 on Build GRCh38
rs772895470 Chr.1: 40301359-40301359 on Build GRCh38
rs115675008 Chr.1: 40301194-40301194 on Build GRCh38
rs113408847 Chr.1: 40301357-40301357 on Build GRCh38
rs773159349 Chr.1: 40301280-40301280 on Build GRCh38
rs866060183 Chr.1: 40301463-40301463 on Build GRCh38
rs201847956 Chr.1: 40301193-40301193 on Build GRCh38
rs750896997 Chr.1: 40301311-40301311 on Build GRCh38
rs1009733110 Chr.1: 40301528-40301528 on Build GRCh38
rs144539022 Chr.1: 40301269-40301269 on Build GRCh38
rs760269057 Chr.1: 40301406-40301406 on Build GRCh38
rs142867960 Chr.1: 40301290-40301290 on Build GRCh38
rs749736976 Chr.1: 40301232-40301232 on Build GRCh38
rs758844390 Chr.1: 40301323-40301323 on Build GRCh38
rs552206262 Chr.1: 40301484-40301484 on Build GRCh38
rs762817478 Chr.1: 40301281-40301281 on Build GRCh38
rs780045313 Chr.1: 40301327-40301327 on Build GRCh38
rs904873014 Chr.1: 40301157-40301157 on Build GRCh38
rs201000437 Chr.1: 40301369-40301369 on Build GRCh38
rs769578991 Chr.1: 40301351-40301351 on Build GRCh38
rs757681519 Chr.1: 40301302-40301302 on Build GRCh38
rs199897562 Chr.1: 40301216-40301216 on Build GRCh38
rs779524161 Chr.1: 40301234-40301234 on Build GRCh38
rs896202757 Chr.1: 40301503-40301503 on Build GRCh38
rs778216645 Chr.1: 40301230-40301230 on Build GRCh38
rs1036771449 Chr.1: 40301219-40301219 on Build GRCh38
rs765050254 Chr.1: 40301272-40301272 on Build GRCh38
rs760921400 Chr.1: 40301186-40301186 on Build GRCh38
rs140041506 Chr.1: 40301271-40301271 on Build GRCh38
rs747178798 Chr.1: 40301267-40301267 on Build GRCh38
rs761675654 Chr.1: 40301198-40301198 on Build GRCh38
rs1056718072 Chr.1: 40301603-40301603 on Build GRCh38
rs764864718 Chr.1: 40301172-40301172 on Build GRCh38
rs775572033 Chr.1: 40301259-40301259 on Build GRCh38
rs1037225008 Chr.1: 40301500-40301501 on Build GRCh38
rs747162216 Chr.1: 40301334-40301334 on Build GRCh38
rs372902486 Chr.1: 40301389-40301389 on Build GRCh38
rs1042535928 Chr.1: 40301538-40301538 on Build GRCh38
rs930979615 Chr.1: 40301537-40301538 on Build GRCh38
rs930549037 Chr.1: 40301333-40301333 on Build GRCh38
rs199850925 Chr.1: 40301300-40301300 on Build GRCh38
rs1002428610 Chr.1: 40301571-40301571 on Build GRCh38
rs754688437 Chr.1: 40301223-40301223 on Build GRCh38
rs569464393 Chr.1: 40301250-40301250 on Build GRCh38
rs767247863 Chr.1: 40301390-40301390 on Build GRCh38
rs752359933 Chr.1: 40301394-40301394 on Build GRCh38
rs919105938 Chr.1: 40301297-40301297 on Build GRCh38
rs150687987 Chr.1: 40301270-40301270 on Build GRCh38
rs1028511698 Chr.1: 40301577-40301577 on Build GRCh38
rs372363712 Chr.1: 40301371-40301371 on Build GRCh38
rs199547057 Chr.1: 40301217-40301217 on Build GRCh38
rs781428852 Chr.1: 40301350-40301350 on Build GRCh38
rs754234353 Chr.1: 40301301-40301301 on Build GRCh38
rs765582256 Chr.1: 40301414-40301414 on Build GRCh38
rs757963762 Chr.1: 40301215-40301215 on Build GRCh38

View Full Product Details