Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41744383-41744889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

SNP Detail

SNP ID Chromosome Location
rs886059412 Chr.4: 41744538-41744538 on Build GRCh38
rs909684800 Chr.4: 41744580-41744580 on Build GRCh38
rs953745494 Chr.4: 41744793-41744793 on Build GRCh38
rs1013070026 Chr.4: 41744407-41744407 on Build GRCh38
rs973671146 Chr.4: 41744839-41744839 on Build GRCh38
rs62412180 Chr.4: 41744460-41744460 on Build GRCh38
rs963831648 Chr.4: 41744419-41744419 on Build GRCh38
rs549166447 Chr.4: 41744759-41744759 on Build GRCh38
rs954074046 Chr.4: 41744479-41744479 on Build GRCh38
rs973454408 Chr.4: 41744597-41744597 on Build GRCh38
rs940975226 Chr.4: 41744498-41744498 on Build GRCh38
rs948957492 Chr.4: 41744697-41744697 on Build GRCh38
rs1044678870 Chr.4: 41744698-41744701 on Build GRCh38
rs1013921778 Chr.4: 41744657-41744657 on Build GRCh38
rs962507653 Chr.4: 41744841-41744841 on Build GRCh38
rs1037946232 Chr.4: 41744512-41744512 on Build GRCh38
rs560506613 Chr.4: 41744483-41744483 on Build GRCh38
rs962054269 Chr.4: 41744799-41744799 on Build GRCh38
rs1000549328 Chr.4: 41744699-41744699 on Build GRCh38
rs1023117234 Chr.4: 41744411-41744411 on Build GRCh38
rs972567482 Chr.4: 41744845-41744846 on Build GRCh38
rs920788258 Chr.4: 41744553-41744553 on Build GRCh38
rs920820822 Chr.4: 41744845-41744845 on Build GRCh38
rs6826373 Chr.4: 41744443-41744443 on Build GRCh38
rs11723860 Chr.4: 41744420-41744420 on Build GRCh38
rs985355555 Chr.4: 41744560-41744560 on Build GRCh38
rs902839590 Chr.4: 41744696-41744696 on Build GRCh38
rs976859318 Chr.4: 41744451-41744451 on Build GRCh38
rs59260453 Chr.4: 41744426-41744426 on Build GRCh38
rs118046131 Chr.4: 41744681-41744681 on Build GRCh38
rs987565912 Chr.4: 41744783-41744783 on Build GRCh38
rs772252596 Chr.4: 41744722-41744722 on Build GRCh38
rs781647693 Chr.4: 41744406-41744406 on Build GRCh38
rs929308592 Chr.4: 41744675-41744675 on Build GRCh38
rs564096488 Chr.4: 41744464-41744464 on Build GRCh38
rs537473339 Chr.4: 41744625-41744625 on Build GRCh38
rs1032871765 Chr.4: 41744715-41744715 on Build GRCh38
rs1055567866 Chr.4: 41744632-41744632 on Build GRCh38
rs941199874 Chr.4: 41744596-41744596 on Build GRCh38
rs180795407 Chr.4: 41744497-41744497 on Build GRCh38
rs184952230 Chr.4: 41744765-41744765 on Build GRCh38
rs560413438 Chr.4: 41744651-41744651 on Build GRCh38
rs1016843942 Chr.4: 41744834-41744834 on Build GRCh38
rs544504353 Chr.4: 41744463-41744463 on Build GRCh38
rs996606230 Chr.4: 41744736-41744736 on Build GRCh38
rs937471360 Chr.4: 41744587-41744587 on Build GRCh38
rs778914258 Chr.4: 41744641-41744641 on Build GRCh38
rs770971831 Chr.4: 41744792-41744792 on Build GRCh38
rs1031428810 Chr.4: 41744427-41744427 on Build GRCh38
rs756699047 Chr.4: 41744435-41744435 on Build GRCh38

View Full Product Details