Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 93757671-93758182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LGI1
Entrez Gene ID
9211
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
UniGene
Hs.533670
Gene Chromosome Location
Chr.10: 93757770-93798174 on Build GRCh38
Transcripts
NM_001308275.1 NM_001308276.1 NM_005097.3 NR_131777.1 XM_017016911.1 XM_017016912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461414 c.10G>A p.E4K Chr.10: 93758154-93758154 on Build GRCh38
6583540 c.28G>T p.G10* Chr.10: 93758172-93758172 on Build GRCh38
3441731 c.13A>T p.R5* Chr.10: 93758157-93758157 on Build GRCh38
7147939 c.10G>C p.E4Q Chr.10: 93758154-93758154 on Build GRCh38
5999338 c.18C>T p.S6S Chr.10: 93758162-93758162 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs924639198 Chr.10: 93758077-93758077 on Build GRCh38
rs534429348 Chr.10: 93758035-93758035 on Build GRCh38
rs1033284841 Chr.10: 93757792-93757792 on Build GRCh38
rs1015408336 Chr.10: 93757920-93757920 on Build GRCh38
rs752138146 Chr.10: 93758132-93758132 on Build GRCh38
rs555527650 Chr.10: 93757724-93757724 on Build GRCh38
rs924588439 Chr.10: 93757757-93757757 on Build GRCh38
rs886047484 Chr.10: 93757841-93757841 on Build GRCh38
rs748209259 Chr.10: 93758156-93758156 on Build GRCh38
rs866303546 Chr.10: 93757948-93757948 on Build GRCh38
rs960320961 Chr.10: 93758079-93758079 on Build GRCh38
rs202204627 Chr.10: 93758145-93758145 on Build GRCh38
rs763835165 Chr.10: 93758119-93758119 on Build GRCh38
rs200602449 Chr.10: 93758138-93758138 on Build GRCh38
rs569225609 Chr.10: 93757848-93757848 on Build GRCh38
rs999536845 Chr.10: 93757754-93757754 on Build GRCh38
rs993673377 Chr.10: 93757748-93757765 on Build GRCh38
rs770920711 Chr.10: 93758063-93758063 on Build GRCh38
rs74150339 Chr.10: 93757865-93757865 on Build GRCh38
rs886770823 Chr.10: 93757892-93757892 on Build GRCh38
rs1044880612 Chr.10: 93757815-93757815 on Build GRCh38
rs879390887 Chr.10: 93757977-93757977 on Build GRCh38
rs995369717 Chr.10: 93757856-93757856 on Build GRCh38
rs919815865 Chr.10: 93757722-93757722 on Build GRCh38
rs914847115 Chr.10: 93757733-93757733 on Build GRCh38
rs963808361 Chr.10: 93757969-93757969 on Build GRCh38
rs949113710 Chr.10: 93757742-93757742 on Build GRCh38
rs758806720 Chr.10: 93758122-93758122 on Build GRCh38
rs1055167121 Chr.10: 93757732-93757732 on Build GRCh38
rs938788682 Chr.10: 93757758-93757758 on Build GRCh38
rs549155243 Chr.10: 93757777-93757777 on Build GRCh38
rs182751947 Chr.10: 93757739-93757739 on Build GRCh38
rs752237435 Chr.10: 93758098-93758098 on Build GRCh38
rs977450179 Chr.10: 93757750-93757750 on Build GRCh38
rs150194814 Chr.10: 93757918-93757918 on Build GRCh38
rs778889740 Chr.10: 93758129-93758129 on Build GRCh38
rs55924739 Chr.10: 93757743-93757743 on Build GRCh38
rs571736665 Chr.10: 93757962-93757962 on Build GRCh38
rs565741397 Chr.10: 93757973-93757973 on Build GRCh38
rs886047485 Chr.10: 93758079-93758079 on Build GRCh38
rs961214748 Chr.10: 93757924-93757924 on Build GRCh38
rs140909783 Chr.10: 93757772-93757772 on Build GRCh38
rs990540455 Chr.10: 93758081-93758081 on Build GRCh38
rs556519003 Chr.10: 93757747-93757747 on Build GRCh38
rs866672066 Chr.10: 93758034-93758034 on Build GRCh38
rs1028239991 Chr.10: 93757858-93757858 on Build GRCh38
rs780231085 Chr.10: 93758128-93758128 on Build GRCh38
rs112058882 Chr.10: 93758004-93758004 on Build GRCh38
rs760462532 Chr.10: 93758109-93758109 on Build GRCh38
rs992802844 Chr.10: 93758085-93758085 on Build GRCh38
rs947881787 Chr.10: 93757800-93757800 on Build GRCh38
rs753475754 Chr.10: 93758121-93758121 on Build GRCh38
rs1008255886 Chr.10: 93757877-93757877 on Build GRCh38

View Full Product Details