Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47467832-47468346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELF1
Entrez Gene ID
10658
Gene Name
CUGBP, Elav-like family member 1
Gene Aliases
BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50
UniGene
Hs.595333
Gene Chromosome Location
Chr.11: 47465937-47565569 on Build GRCh38
Transcripts
NM_001025596.2 NM_001172639.1 NM_001172640.1 NM_006560.3 NM_198700.2 XM_011519848.2 XM_011519850.2 XM_011519852.2 XM_011519854.1 XM_011519855.1 XM_011519856.1 XM_011519857.1 XM_011519858.1 XM_011519859.1 XM_017017101.1 XM_017017102.1 XM_017017103.1 XM_017017104.1 XM_017017105.1 XM_017017106.1 XM_017017107.1 XM_017017108.1 XM_017017109.1 XM_017017110.1 XM_017017111.1 XM_017017112.1 XM_017017113.1 XM_017017114.1 XM_017017115.1 XM_017017116.1 XM_017017117.1 XM_017017118.1 XM_017017119.1 XM_017017120.1 XM_017017121.1 XM_017017122.1 XM_017017123.1 XM_017017124.1 XM_017017125.1 XM_017017126.1 XM_017017127.1 XM_017017128.1 XM_017017129.1 XM_017017130.1 XM_017017131.1 XM_017017132.1 XM_017017133.1 XM_017017134.1 XM_017017135.1

SNP Detail

SNP ID Chromosome Location
rs1035257311 Chr.11: 47467902-47467902 on Build GRCh38
rs976589433 Chr.11: 47468125-47468125 on Build GRCh38
rs763614856 Chr.11: 47467943-47467943 on Build GRCh38
rs565298127 Chr.11: 47468194-47468194 on Build GRCh38
rs912946090 Chr.11: 47468134-47468134 on Build GRCh38
rs886134989 Chr.11: 47468150-47468150 on Build GRCh38
rs545624506 Chr.11: 47468170-47468170 on Build GRCh38
rs372536437 Chr.11: 47467927-47467927 on Build GRCh38
rs1023846662 Chr.11: 47468229-47468229 on Build GRCh38
rs896067101 Chr.11: 47468205-47468205 on Build GRCh38
rs531234987 Chr.11: 47468233-47468233 on Build GRCh38
rs986778487 Chr.11: 47468316-47468316 on Build GRCh38
rs911830301 Chr.11: 47468087-47468087 on Build GRCh38
rs879679788 Chr.11: 47468213-47468218 on Build GRCh38
rs557145148 Chr.11: 47467878-47467881 on Build GRCh38
rs745516706 Chr.11: 47468084-47468084 on Build GRCh38
rs1014930787 Chr.11: 47467928-47467928 on Build GRCh38
rs199995378 Chr.11: 47468227-47468228 on Build GRCh38
rs369566397 Chr.11: 47468230-47468231 on Build GRCh38
rs953533779 Chr.11: 47468029-47468029 on Build GRCh38
rs577523524 Chr.11: 47467938-47467938 on Build GRCh38
rs979920763 Chr.11: 47468035-47468035 on Build GRCh38
rs1028178766 Chr.11: 47468304-47468304 on Build GRCh38
rs1031482685 Chr.11: 47468265-47468265 on Build GRCh38
rs985377706 Chr.11: 47468088-47468090 on Build GRCh38
rs935313545 Chr.11: 47468059-47468059 on Build GRCh38
rs968160918 Chr.11: 47468018-47468018 on Build GRCh38
rs1014392035 Chr.11: 47467905-47467905 on Build GRCh38
rs1005168995 Chr.11: 47467864-47467864 on Build GRCh38
rs895964594 Chr.11: 47467893-47467893 on Build GRCh38
rs561549938 Chr.11: 47468300-47468300 on Build GRCh38
rs973500231 Chr.11: 47467962-47467962 on Build GRCh38
rs370403413 Chr.11: 47468297-47468297 on Build GRCh38
rs1000984552 Chr.11: 47468264-47468264 on Build GRCh38
rs528422329 Chr.11: 47468278-47468279 on Build GRCh38
rs960924584 Chr.11: 47467908-47467908 on Build GRCh38
rs111593711 Chr.11: 47468165-47468165 on Build GRCh38
rs1036488890 Chr.11: 47468261-47468262 on Build GRCh38
rs896748308 Chr.11: 47468168-47468168 on Build GRCh38
rs944598788 Chr.11: 47468090-47468090 on Build GRCh38
rs968612373 Chr.11: 47468251-47468251 on Build GRCh38
rs573270337 Chr.11: 47468093-47468093 on Build GRCh38
rs1023774428 Chr.11: 47467996-47467996 on Build GRCh38
rs930449478 Chr.11: 47468099-47468099 on Build GRCh38
rs559428581 Chr.11: 47468158-47468158 on Build GRCh38
rs1052657 Chr.11: 47468148-47468148 on Build GRCh38
rs190844690 Chr.11: 47468013-47468013 on Build GRCh38
rs377227389 Chr.11: 47468298-47468298 on Build GRCh38
rs185936690 Chr.11: 47467979-47467979 on Build GRCh38
rs988770978 Chr.11: 47468077-47468077 on Build GRCh38
rs1004974070 Chr.11: 47468161-47468161 on Build GRCh38
rs1038852951 Chr.11: 47468092-47468092 on Build GRCh38
rs923997009 Chr.11: 47468051-47468051 on Build GRCh38

View Full Product Details