Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74919365-74919877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521966 c.1132G>A p.D378N Chr.17: 74919704-74919704 on Build GRCh38
5456645 c.1361C>T p.P454L Chr.17: 74919475-74919475 on Build GRCh38
6556658 c.1114C>A p.L372M Chr.17: 74919722-74919722 on Build GRCh38
4525401 c.1329G>A p.V443V Chr.17: 74919507-74919507 on Build GRCh38
1385864 c.1283G>A p.S428N Chr.17: 74919553-74919553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576500685 Chr.17: 74919687-74919687 on Build GRCh38
rs116872904 Chr.17: 74919637-74919637 on Build GRCh38
rs569032124 Chr.17: 74919684-74919684 on Build GRCh38
rs971548417 Chr.17: 74919490-74919490 on Build GRCh38
rs1023348230 Chr.17: 74919770-74919770 on Build GRCh38
rs267605044 Chr.17: 74919700-74919700 on Build GRCh38
rs747285177 Chr.17: 74919475-74919475 on Build GRCh38
rs372360601 Chr.17: 74919566-74919566 on Build GRCh38
rs1023210382 Chr.17: 74919830-74919830 on Build GRCh38
rs765888502 Chr.17: 74919417-74919417 on Build GRCh38
rs774725935 Chr.17: 74919672-74919672 on Build GRCh38
rs746578014 Chr.17: 74919788-74919788 on Build GRCh38
rs760743915 Chr.17: 74919409-74919409 on Build GRCh38
rs778970061 Chr.17: 74919842-74919842 on Build GRCh38
rs746392682 Chr.17: 74919760-74919760 on Build GRCh38
rs997638798 Chr.17: 74919628-74919628 on Build GRCh38
rs375297515 Chr.17: 74919451-74919451 on Build GRCh38
rs762354629 Chr.17: 74919718-74919718 on Build GRCh38
rs776149260 Chr.17: 74919797-74919797 on Build GRCh38
rs142291012 Chr.17: 74919782-74919782 on Build GRCh38
rs757991416 Chr.17: 74919423-74919423 on Build GRCh38
rs766484391 Chr.17: 74919815-74919815 on Build GRCh38
rs755838297 Chr.17: 74919500-74919501 on Build GRCh38
rs776814552 Chr.17: 74919411-74919411 on Build GRCh38
rs1016117261 Chr.17: 74919708-74919708 on Build GRCh38
rs751073735 Chr.17: 74919510-74919510 on Build GRCh38
rs200613072 Chr.17: 74919754-74919754 on Build GRCh38
rs201359669 Chr.17: 74919496-74919496 on Build GRCh38
rs886043626 Chr.17: 74919525-74919525 on Build GRCh38
rs868106772 Chr.17: 74919635-74919635 on Build GRCh38
rs761467615 Chr.17: 74919803-74919803 on Build GRCh38
rs778876458 Chr.17: 74919551-74919551 on Build GRCh38
rs754536324 Chr.17: 74919435-74919435 on Build GRCh38
rs117489945 Chr.17: 74919611-74919611 on Build GRCh38
rs748416580 Chr.17: 74919634-74919634 on Build GRCh38
rs373882524 Chr.17: 74919434-74919434 on Build GRCh38
rs772606877 Chr.17: 74919773-74919773 on Build GRCh38
rs991658805 Chr.17: 74919449-74919449 on Build GRCh38
rs886265451 Chr.17: 74919678-74919678 on Build GRCh38
rs368110384 Chr.17: 74919557-74919557 on Build GRCh38
rs368389700 Chr.17: 74919436-74919436 on Build GRCh38
rs538983393 Chr.17: 74919743-74919743 on Build GRCh38
rs867140684 Chr.17: 74919683-74919683 on Build GRCh38
rs748994272 Chr.17: 74919762-74919776 on Build GRCh38
rs778771485 Chr.17: 74919621-74919621 on Build GRCh38
rs779288250 Chr.17: 74919759-74919759 on Build GRCh38
rs369036189 Chr.17: 74919662-74919662 on Build GRCh38
rs766986350 Chr.17: 74919597-74919597 on Build GRCh38
rs750965243 Chr.17: 74919420-74919420 on Build GRCh38
rs750367080 Chr.17: 74919629-74919629 on Build GRCh38
rs746328214 Chr.17: 74919674-74919674 on Build GRCh38
rs139897506 Chr.17: 74919578-74919578 on Build GRCh38
rs754029450 Chr.17: 74919840-74919840 on Build GRCh38
rs762322942 Chr.17: 74919482-74919482 on Build GRCh38
rs137871047 Chr.17: 74919853-74919853 on Build GRCh38
rs914830331 Chr.17: 74919503-74919503 on Build GRCh38
rs955918806 Chr.17: 74919522-74919522 on Build GRCh38
rs764553015 Chr.17: 74919410-74919410 on Build GRCh38
rs397517925 Chr.17: 74919463-74919463 on Build GRCh38
rs770166095 Chr.17: 74919767-74919767 on Build GRCh38
rs755013643 Chr.17: 74919630-74919630 on Build GRCh38
rs763348976 Chr.17: 74919586-74919586 on Build GRCh38
rs148479646 Chr.17: 74919751-74919751 on Build GRCh38
rs199724052 Chr.17: 74919666-74919666 on Build GRCh38
rs767134204 Chr.17: 74919519-74919519 on Build GRCh38
rs962741334 Chr.17: 74919435-74919436 on Build GRCh38
rs924339909 Chr.17: 74919505-74919505 on Build GRCh38
rs781625717 Chr.17: 74919563-74919563 on Build GRCh38
rs762822503 Chr.17: 74919814-74919814 on Build GRCh38
rs765736586 Chr.17: 74919431-74919431 on Build GRCh38
rs373170645 Chr.17: 74919845-74919845 on Build GRCh38
rs551386496 Chr.17: 74919447-74919447 on Build GRCh38
rs369623755 Chr.17: 74919757-74919757 on Build GRCh38
rs777486419 Chr.17: 74919539-74919539 on Build GRCh38
rs751540523 Chr.17: 74919817-74919817 on Build GRCh38
rs781376716 Chr.17: 74919633-74919633 on Build GRCh38
rs777537471 Chr.17: 74919603-74919605 on Build GRCh38
rs989804696 Chr.17: 74919523-74919523 on Build GRCh38
rs756268562 Chr.17: 74919821-74919821 on Build GRCh38
rs866075782 Chr.17: 74919839-74919839 on Build GRCh38
rs764976687 Chr.17: 74919811-74919811 on Build GRCh38
rs943263343 Chr.17: 74919555-74919555 on Build GRCh38
rs763508231 Chr.17: 74919499-74919499 on Build GRCh38
rs768147159 Chr.17: 74919790-74919790 on Build GRCh38
rs753794521 Chr.17: 74919545-74919545 on Build GRCh38
rs780964009 Chr.17: 74919697-74919697 on Build GRCh38
rs970523771 Chr.17: 74919437-74919437 on Build GRCh38
rs757101131 Chr.17: 74919550-74919550 on Build GRCh38
rs1004713388 Chr.17: 74919696-74919696 on Build GRCh38
rs752513148 Chr.17: 74919493-74919494 on Build GRCh38
rs559001977 Chr.17: 74919750-74919750 on Build GRCh38
rs371983289 Chr.17: 74919695-74919695 on Build GRCh38
rs1004192113 Chr.17: 74919432-74919432 on Build GRCh38
rs151242039 Chr.17: 74919824-74919824 on Build GRCh38
rs879066143 Chr.17: 74919844-74919844 on Build GRCh38
rs759065977 Chr.17: 74919518-74919518 on Build GRCh38
rs762249466 Chr.17: 74919416-74919416 on Build GRCh38
rs761576510 Chr.17: 74919690-74919690 on Build GRCh38
rs775795179 Chr.17: 74919404-74919404 on Build GRCh38
rs757051357 Chr.17: 74919615-74919615 on Build GRCh38
rs773872479 Chr.17: 74919497-74919497 on Build GRCh38
rs752343151 Chr.17: 74919529-74919529 on Build GRCh38
rs776968656 Chr.17: 74919481-74919481 on Build GRCh38
rs773103971 Chr.17: 74919812-74919812 on Build GRCh38
rs775952116 Chr.17: 74919779-74919779 on Build GRCh38
rs775382564 Chr.17: 74919465-74919465 on Build GRCh38
rs758319467 Chr.17: 74919704-74919704 on Build GRCh38
rs374693956 Chr.17: 74919577-74919577 on Build GRCh38
rs780954300 Chr.17: 74919428-74919429 on Build GRCh38
rs146287064 Chr.17: 74919657-74919657 on Build GRCh38
rs774909603 Chr.17: 74919596-74919596 on Build GRCh38
rs771392412 Chr.17: 74919593-74919593 on Build GRCh38
rs748326974 Chr.17: 74919564-74919564 on Build GRCh38
rs760271310 Chr.17: 74919606-74919606 on Build GRCh38
rs765949002 Chr.17: 74919500-74919500 on Build GRCh38
rs141854343 Chr.17: 74919645-74919645 on Build GRCh38
rs375322161 Chr.17: 74919749-74919749 on Build GRCh38
rs755890365 Chr.17: 74919438-74919438 on Build GRCh38
rs768787249 Chr.17: 74919401-74919401 on Build GRCh38
rs372580540 Chr.17: 74919478-74919478 on Build GRCh38
rs776330616 Chr.17: 74919688-74919688 on Build GRCh38
rs963596546 Chr.17: 74919719-74919719 on Build GRCh38
rs752359635 Chr.17: 74919604-74919604 on Build GRCh38
rs898486902 Chr.17: 74919792-74919792 on Build GRCh38
rs866052000 Chr.17: 74919622-74919622 on Build GRCh38
rs113503320 Chr.17: 74919787-74919787 on Build GRCh38
rs755908331 Chr.17: 74919530-74919530 on Build GRCh38

View Full Product Details