Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 107649798-107650344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRPS1
Entrez Gene ID
5631
Gene Name
phosphoribosyl pyrophosphate synthetase 1
Gene Aliases
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI
UniGene
Hs.56
Gene Chromosome Location
Chr.X: 107628424-107651026 on Build GRCh38
Transcripts
NM_001204402.1 NM_002764.3

SNP Detail

SNP ID Chromosome Location
rs773323331 Chr.X: 107649908-107649908 on Build GRCh38
rs185553780 Chr.X: 107649863-107649863 on Build GRCh38
rs751468763 Chr.X: 107649990-107649990 on Build GRCh38
rs768298456 Chr.X: 107650071-107650071 on Build GRCh38
rs760711543 Chr.X: 107649909-107649909 on Build GRCh38
rs587781263 Chr.X: 107650000-107650000 on Build GRCh38
rs1126445 Chr.X: 107650169-107650169 on Build GRCh38
rs970838701 Chr.X: 107649923-107649923 on Build GRCh38
rs777440100 Chr.X: 107650190-107650190 on Build GRCh38
rs758809582 Chr.X: 107650176-107650176 on Build GRCh38
rs918359560 Chr.X: 107650120-107650120 on Build GRCh38
rs763915105 Chr.X: 107649974-107649974 on Build GRCh38
rs142438413 Chr.X: 107649821-107649821 on Build GRCh38
rs375588087 Chr.X: 107649904-107649904 on Build GRCh38
rs756026687 Chr.X: 107650152-107650152 on Build GRCh38
rs371265973 Chr.X: 107650198-107650198 on Build GRCh38
rs766514496 Chr.X: 107649926-107649926 on Build GRCh38
rs752535865 Chr.X: 107650226-107650227 on Build GRCh38
rs779558146 Chr.X: 107650054-107650054 on Build GRCh38
rs758574318 Chr.X: 107650285-107650287 on Build GRCh38
rs576933222 Chr.X: 107650210-107650210 on Build GRCh38
rs202233161 Chr.X: 107649951-107649951 on Build GRCh38
rs952452306 Chr.X: 107650133-107650133 on Build GRCh38
rs748877902 Chr.X: 107650068-107650068 on Build GRCh38
rs867662574 Chr.X: 107649964-107649964 on Build GRCh38
rs754068855 Chr.X: 107649945-107649945 on Build GRCh38
rs180177154 Chr.X: 107649991-107649991 on Build GRCh38
rs1057515726 Chr.X: 107650120-107650120 on Build GRCh38
rs755539554 Chr.X: 107650047-107650047 on Build GRCh38
rs757207081 Chr.X: 107649999-107649999 on Build GRCh38
rs747334780 Chr.X: 107650191-107650191 on Build GRCh38
rs752673062 Chr.X: 107650114-107650114 on Build GRCh38
rs180177153 Chr.X: 107649944-107649944 on Build GRCh38
rs942898095 Chr.X: 107650187-107650187 on Build GRCh38
rs767972803 Chr.X: 107649900-107649900 on Build GRCh38
rs147731055 Chr.X: 107650017-107650017 on Build GRCh38

View Full Product Details