Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 75008583-75009147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B2
Entrez Gene ID
8892
Gene Name
eukaryotic translation initiation factor 2B subunit beta
Gene Aliases
EIF-2Bbeta, EIF2B
UniGene
Hs.409137
Gene Chromosome Location
Chr.14: 75002909-75009591 on Build GRCh38
Transcripts
NM_014239.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6266771 c.946G>T p.V316F Chr.14: 75009078-75009078 on Build GRCh38
6652713 c.921C>T p.S307S Chr.14: 75009053-75009053 on Build GRCh38
957894 c.922G>A p.V308M Chr.14: 75009054-75009054 on Build GRCh38
4788678 c.927T>C p.H309H Chr.14: 75009059-75009059 on Build GRCh38
6652712 c.899-2A>G p.? Chr.14: 75009029-75009029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs539011059 Chr.14: 75008920-75008920 on Build GRCh38
rs117225020 Chr.14: 75008863-75008863 on Build GRCh38
rs1036228873 Chr.14: 75009016-75009016 on Build GRCh38
rs997254799 Chr.14: 75008661-75008661 on Build GRCh38
rs969006514 Chr.14: 75008796-75008796 on Build GRCh38
rs1004322147 Chr.14: 75008774-75008774 on Build GRCh38
rs765712931 Chr.14: 75009105-75009105 on Build GRCh38
rs888646929 Chr.14: 75008730-75008730 on Build GRCh38
rs767183422 Chr.14: 75009062-75009062 on Build GRCh38
rs1031885026 Chr.14: 75008827-75008828 on Build GRCh38
rs776134292 Chr.14: 75008642-75008642 on Build GRCh38
rs113994018 Chr.14: 75009042-75009042 on Build GRCh38
rs747445417 Chr.14: 75008687-75008687 on Build GRCh38
rs756121992 Chr.14: 75009018-75009018 on Build GRCh38
rs915981637 Chr.14: 75008911-75008911 on Build GRCh38
rs948784270 Chr.14: 75008921-75008921 on Build GRCh38
rs559338892 Chr.14: 75008930-75008930 on Build GRCh38
rs768467979 Chr.14: 75009053-75009053 on Build GRCh38
rs762563184 Chr.14: 75009043-75009045 on Build GRCh38
rs759440505 Chr.14: 75008956-75008956 on Build GRCh38
rs773407374 Chr.14: 75009077-75009077 on Build GRCh38
rs570479314 Chr.14: 75008833-75008833 on Build GRCh38
rs762379190 Chr.14: 75008807-75008807 on Build GRCh38
rs772125418 Chr.14: 75009074-75009074 on Build GRCh38
rs113994020 Chr.14: 75009118-75009118 on Build GRCh38
rs372548739 Chr.14: 75009054-75009054 on Build GRCh38
rs761055346 Chr.14: 75009078-75009078 on Build GRCh38
rs773976168 Chr.14: 75008927-75008927 on Build GRCh38
rs780910747 Chr.14: 75008993-75008993 on Build GRCh38
rs577844198 Chr.14: 75009111-75009111 on Build GRCh38
rs867623753 Chr.14: 75008964-75008964 on Build GRCh38
rs538441859 Chr.14: 75008943-75008943 on Build GRCh38
rs749059127 Chr.14: 75009033-75009033 on Build GRCh38
rs992943741 Chr.14: 75008897-75008897 on Build GRCh38
rs921537312 Chr.14: 75008689-75008689 on Build GRCh38
rs969334792 Chr.14: 75008922-75008922 on Build GRCh38
rs757439974 Chr.14: 75008952-75008952 on Build GRCh38
rs759914845 Chr.14: 75009092-75009092 on Build GRCh38
rs976783743 Chr.14: 75008990-75008990 on Build GRCh38
rs1041416714 Chr.14: 75008663-75008663 on Build GRCh38
rs957854731 Chr.14: 75008603-75008603 on Build GRCh38
rs138817058 Chr.14: 75009059-75009059 on Build GRCh38
rs145117455 Chr.14: 75009114-75009114 on Build GRCh38
rs909833252 Chr.14: 75008941-75008941 on Build GRCh38
rs35607382 Chr.14: 75008741-75008742 on Build GRCh38
rs772735678 Chr.14: 75008802-75008802 on Build GRCh38
rs112186469 Chr.14: 75008720-75008720 on Build GRCh38
rs575999461 Chr.14: 75008934-75008934 on Build GRCh38
rs142254655 Chr.14: 75008975-75008975 on Build GRCh38
rs745792330 Chr.14: 75009006-75009006 on Build GRCh38
rs988253405 Chr.14: 75008606-75008606 on Build GRCh38
rs1013264049 Chr.14: 75008813-75008813 on Build GRCh38
rs1030024787 Chr.14: 75008664-75008664 on Build GRCh38
rs932868051 Chr.14: 75008717-75008717 on Build GRCh38
rs987950473 Chr.14: 75008951-75008951 on Build GRCh38
rs796126953 Chr.14: 75008937-75008937 on Build GRCh38
rs375585987 Chr.14: 75009116-75009116 on Build GRCh38
rs200849948 Chr.14: 75009112-75009112 on Build GRCh38
rs944072007 Chr.14: 75008660-75008660 on Build GRCh38
rs766123244 Chr.14: 75009096-75009098 on Build GRCh38
rs78900727 Chr.14: 75009020-75009020 on Build GRCh38
rs104894426 Chr.14: 75009079-75009079 on Build GRCh38
rs888884716 Chr.14: 75009080-75009080 on Build GRCh38
rs953187124 Chr.14: 75008674-75008674 on Build GRCh38
rs1021680119 Chr.14: 75008792-75008792 on Build GRCh38
rs1021051962 Chr.14: 75008974-75008974 on Build GRCh38
rs777036076 Chr.14: 75009086-75009086 on Build GRCh38
rs749995021 Chr.14: 75009124-75009124 on Build GRCh38

View Full Product Details