Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74920124-74920671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7243366 c.591C>T p.S197S Chr.17: 74920245-74920245 on Build GRCh38
1303319 c.515G>C p.R172P Chr.17: 74920321-74920321 on Build GRCh38
5583009 c.502G>A p.E168K Chr.17: 74920334-74920334 on Build GRCh38
7127916 c.693C>T p.F231F Chr.17: 74920143-74920143 on Build GRCh38
7138382 c.206delT p.L69fs*26 Chr.17: 74920630-74920630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200082225 Chr.17: 74920403-74920403 on Build GRCh38
rs750109903 Chr.17: 74920237-74920237 on Build GRCh38
rs147972772 Chr.17: 74920305-74920305 on Build GRCh38
rs745450960 Chr.17: 74920572-74920572 on Build GRCh38
rs199941325 Chr.17: 74920322-74920322 on Build GRCh38
rs773698651 Chr.17: 74920380-74920380 on Build GRCh38
rs747169902 Chr.17: 74920347-74920347 on Build GRCh38
rs759026779 Chr.17: 74920457-74920457 on Build GRCh38
rs373704838 Chr.17: 74920254-74920254 on Build GRCh38
rs727503713 Chr.17: 74920242-74920242 on Build GRCh38
rs763891710 Chr.17: 74920496-74920496 on Build GRCh38
rs1003052325 Chr.17: 74920171-74920171 on Build GRCh38
rs138590256 Chr.17: 74920356-74920356 on Build GRCh38
rs769161031 Chr.17: 74920349-74920349 on Build GRCh38
rs771517713 Chr.17: 74920556-74920556 on Build GRCh38
rs781540158 Chr.17: 74920418-74920418 on Build GRCh38
rs745992000 Chr.17: 74920336-74920336 on Build GRCh38
rs111033466 Chr.17: 74920412-74920412 on Build GRCh38
rs779722969 Chr.17: 74920246-74920246 on Build GRCh38
rs772186029 Chr.17: 74920339-74920339 on Build GRCh38
rs587776546 Chr.17: 74920441-74920442 on Build GRCh38
rs764867252 Chr.17: 74920318-74920318 on Build GRCh38
rs201822644 Chr.17: 74920377-74920377 on Build GRCh38
rs886053390 Chr.17: 74920488-74920488 on Build GRCh38
rs200786476 Chr.17: 74920316-74920316 on Build GRCh38
rs574697331 Chr.17: 74920328-74920328 on Build GRCh38
rs754767231 Chr.17: 74920252-74920252 on Build GRCh38
rs549973959 Chr.17: 74920607-74920607 on Build GRCh38
rs765050197 Chr.17: 74920517-74920517 on Build GRCh38
rs374630813 Chr.17: 74920326-74920326 on Build GRCh38
rs762274233 Chr.17: 74920179-74920181 on Build GRCh38
rs954934463 Chr.17: 74920462-74920462 on Build GRCh38
rs774403004 Chr.17: 74920189-74920189 on Build GRCh38
rs770249797 Chr.17: 74920357-74920357 on Build GRCh38
rs748592728 Chr.17: 74920425-74920425 on Build GRCh38
rs776540260 Chr.17: 74920159-74920159 on Build GRCh38
rs1010293771 Chr.17: 74920393-74920393 on Build GRCh38
rs928656346 Chr.17: 74920645-74920645 on Build GRCh38
rs376527217 Chr.17: 74920358-74920358 on Build GRCh38
rs1012134220 Chr.17: 74920354-74920354 on Build GRCh38
rs751196707 Chr.17: 74920368-74920368 on Build GRCh38
rs397517927 Chr.17: 74920585-74920585 on Build GRCh38
rs963261823 Chr.17: 74920554-74920554 on Build GRCh38
rs775135769 Chr.17: 74920587-74920587 on Build GRCh38
rs759149374 Chr.17: 74920373-74920373 on Build GRCh38
rs141791769 Chr.17: 74920400-74920400 on Build GRCh38
rs201644674 Chr.17: 74920270-74920270 on Build GRCh38
rs149529031 Chr.17: 74920526-74920526 on Build GRCh38
rs111033465 Chr.17: 74920448-74920448 on Build GRCh38
rs573510359 Chr.17: 74920210-74920210 on Build GRCh38
rs755288377 Chr.17: 74920417-74920417 on Build GRCh38
rs1021385697 Chr.17: 74920380-74920380 on Build GRCh38
rs530264162 Chr.17: 74920160-74920160 on Build GRCh38
rs201525855 Chr.17: 74920158-74920158 on Build GRCh38
rs148496763 Chr.17: 74920578-74920578 on Build GRCh38
rs1021745794 Chr.17: 74920407-74920407 on Build GRCh38
rs374579438 Chr.17: 74920297-74920297 on Build GRCh38
rs201866631 Chr.17: 74920325-74920325 on Build GRCh38
rs995800790 Chr.17: 74920433-74920433 on Build GRCh38
rs774827536 Chr.17: 74920570-74920570 on Build GRCh38
rs751735275 Chr.17: 74920524-74920524 on Build GRCh38
rs140951373 Chr.17: 74920447-74920447 on Build GRCh38
rs756087370 Chr.17: 74920333-74920333 on Build GRCh38
rs756461951 Chr.17: 74920541-74920541 on Build GRCh38
rs370887019 Chr.17: 74920431-74920431 on Build GRCh38
rs375077331 Chr.17: 74920451-74920451 on Build GRCh38
rs727504890 Chr.17: 74920229-74920229 on Build GRCh38
rs775117744 Chr.17: 74920492-74920492 on Build GRCh38
rs974350317 Chr.17: 74920256-74920256 on Build GRCh38
rs769073148 Chr.17: 74920291-74920291 on Build GRCh38
rs751372463 Chr.17: 74920247-74920247 on Build GRCh38
rs920379501 Chr.17: 74920547-74920547 on Build GRCh38
rs143458197 Chr.17: 74920235-74920235 on Build GRCh38
rs201123735 Chr.17: 74920273-74920273 on Build GRCh38
rs771022122 Chr.17: 74920255-74920255 on Build GRCh38
rs368407264 Chr.17: 74920574-74920574 on Build GRCh38
rs764428038 Chr.17: 74920321-74920321 on Build GRCh38
rs765860407 Chr.17: 74920362-74920362 on Build GRCh38
rs754390030 Chr.17: 74920209-74920209 on Build GRCh38
rs886043212 Chr.17: 74920314-74920314 on Build GRCh38
rs770350814 Chr.17: 74920200-74920201 on Build GRCh38
rs202045256 Chr.17: 74920243-74920243 on Build GRCh38
rs770131812 Chr.17: 74920427-74920427 on Build GRCh38
rs759630941 Chr.17: 74920182-74920182 on Build GRCh38
rs756830882 Chr.17: 74920218-74920218 on Build GRCh38
rs761010255 Chr.17: 74920284-74920284 on Build GRCh38
rs938957013 Chr.17: 74920337-74920337 on Build GRCh38
rs144686462 Chr.17: 74920575-74920575 on Build GRCh38
rs374260443 Chr.17: 74920536-74920536 on Build GRCh38
rs760325139 Chr.17: 74920588-74920588 on Build GRCh38
rs749586302 Chr.17: 74920555-74920555 on Build GRCh38
rs774571563 Chr.17: 74920181-74920181 on Build GRCh38
rs775758795 Chr.17: 74920187-74920187 on Build GRCh38
rs760465755 Chr.17: 74920493-74920493 on Build GRCh38
rs199714230 Chr.17: 74920215-74920215 on Build GRCh38
rs141688757 Chr.17: 74920335-74920335 on Build GRCh38
rs907735153 Chr.17: 74920302-74920302 on Build GRCh38
rs778163212 Chr.17: 74920544-74920544 on Build GRCh38
rs767955984 Chr.17: 74920186-74920186 on Build GRCh38
rs780916600 Chr.17: 74920327-74920327 on Build GRCh38
rs748325007 Chr.17: 74920537-74920537 on Build GRCh38
rs558890451 Chr.17: 74920281-74920281 on Build GRCh38
rs202005955 Chr.17: 74920402-74920402 on Build GRCh38
rs370027457 Chr.17: 74920155-74920155 on Build GRCh38
rs773610870 Chr.17: 74920441-74920441 on Build GRCh38
rs200197601 Chr.17: 74920201-74920201 on Build GRCh38
rs761488604 Chr.17: 74920626-74920626 on Build GRCh38
rs730880268 Chr.17: 74920649-74920650 on Build GRCh38
rs938661179 Chr.17: 74920652-74920652 on Build GRCh38
rs947865484 Chr.17: 74920553-74920553 on Build GRCh38
rs1044385908 Chr.17: 74920331-74920331 on Build GRCh38
rs757174476 Chr.17: 74920516-74920516 on Build GRCh38
rs747261903 Chr.17: 74920415-74920415 on Build GRCh38
rs200345990 Chr.17: 74920605-74920605 on Build GRCh38
rs980182737 Chr.17: 74920593-74920593 on Build GRCh38
rs561119699 Chr.17: 74920476-74920476 on Build GRCh38
rs34854210 Chr.17: 74920334-74920335 on Build GRCh38
rs144633924 Chr.17: 74920345-74920345 on Build GRCh38
rs753740036 Chr.17: 74920505-74920505 on Build GRCh38
rs770933979 Chr.17: 74920168-74920168 on Build GRCh38

View Full Product Details