Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74916031-74916567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USH1G
Entrez Gene ID
124590
Gene Name
USH1 protein network component sans
Gene Aliases
ANKS4A, SANS
UniGene
Hs.376688
Gene Chromosome Location
Chr.17: 74916083-74923263 on Build GRCh38
Transcripts
NM_001282489.2 NM_173477.4 XM_011524296.2

SNP Detail

SNP ID Chromosome Location
rs545706385 Chr.17: 74916207-74916207 on Build GRCh38
rs986965905 Chr.17: 74916221-74916221 on Build GRCh38
rs769169741 Chr.17: 74916364-74916364 on Build GRCh38
rs113905467 Chr.17: 74916083-74916083 on Build GRCh38
rs980965294 Chr.17: 74916216-74916216 on Build GRCh38
rs570830006 Chr.17: 74916483-74916483 on Build GRCh38
rs992304430 Chr.17: 74916494-74916494 on Build GRCh38
rs550526184 Chr.17: 74916481-74916481 on Build GRCh38
rs896617891 Chr.17: 74916329-74916329 on Build GRCh38
rs904126534 Chr.17: 74916344-74916344 on Build GRCh38
rs1009783144 Chr.17: 74916112-74916112 on Build GRCh38
rs74943260 Chr.17: 74916252-74916252 on Build GRCh38
rs945077004 Chr.17: 74916517-74916517 on Build GRCh38
rs757488031 Chr.17: 74916082-74916082 on Build GRCh38
rs939528955 Chr.17: 74916229-74916229 on Build GRCh38
rs1056256261 Chr.17: 74916241-74916241 on Build GRCh38
rs997129784 Chr.17: 74916318-74916318 on Build GRCh38
rs952075985 Chr.17: 74916442-74916442 on Build GRCh38
rs930641698 Chr.17: 74916308-74916308 on Build GRCh38
rs1043386378 Chr.17: 74916254-74916254 on Build GRCh38
rs1028543289 Chr.17: 74916086-74916086 on Build GRCh38
rs8067775 Chr.17: 74916417-74916417 on Build GRCh38
rs961503528 Chr.17: 74916255-74916255 on Build GRCh38
rs559481575 Chr.17: 74916248-74916248 on Build GRCh38
rs993467204 Chr.17: 74916398-74916398 on Build GRCh38
rs886053383 Chr.17: 74916341-74916341 on Build GRCh38
rs1049073936 Chr.17: 74916314-74916314 on Build GRCh38
rs886053382 Chr.17: 74916167-74916168 on Build GRCh38
rs913636146 Chr.17: 74916251-74916251 on Build GRCh38
rs138923906 Chr.17: 74916368-74916368 on Build GRCh38
rs926821931 Chr.17: 74916217-74916217 on Build GRCh38
rs769165276 Chr.17: 74916539-74916539 on Build GRCh38
rs889470597 Chr.17: 74916416-74916416 on Build GRCh38
rs924981787 Chr.17: 74916541-74916541 on Build GRCh38
rs953492455 Chr.17: 74916102-74916102 on Build GRCh38
rs886053381 Chr.17: 74916086-74916086 on Build GRCh38
rs1009436719 Chr.17: 74916125-74916125 on Build GRCh38
rs770108840 Chr.17: 74916141-74916141 on Build GRCh38
rs368769683 Chr.17: 74916238-74916238 on Build GRCh38
rs903778854 Chr.17: 74916271-74916271 on Build GRCh38
rs962040930 Chr.17: 74916162-74916162 on Build GRCh38
rs749721429 Chr.17: 74916206-74916206 on Build GRCh38
rs981462825 Chr.17: 74916444-74916444 on Build GRCh38
rs1049665070 Chr.17: 74916324-74916324 on Build GRCh38
rs919876587 Chr.17: 74916278-74916278 on Build GRCh38
rs554481437 Chr.17: 74916340-74916340 on Build GRCh38
rs967699460 Chr.17: 74916164-74916164 on Build GRCh38
rs913667139 Chr.17: 74916514-74916514 on Build GRCh38
rs890980801 Chr.17: 74916328-74916328 on Build GRCh38
rs746234262 Chr.17: 74916085-74916085 on Build GRCh38
rs1042658077 Chr.17: 74916341-74916341 on Build GRCh38
rs886053384 Chr.17: 74916354-74916354 on Build GRCh38
rs1019003031 Chr.17: 74916354-74916354 on Build GRCh38
rs1027966300 Chr.17: 74916412-74916412 on Build GRCh38
rs1568448 Chr.17: 74916173-74916173 on Build GRCh38
rs760694751 Chr.17: 74916506-74916506 on Build GRCh38
rs1000517704 Chr.17: 74916358-74916358 on Build GRCh38
rs374499352 Chr.17: 74916433-74916441 on Build GRCh38
rs868650355 Chr.17: 74916151-74916151 on Build GRCh38

View Full Product Details