Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112418691-112418881 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 NM_001320141.1 XM_017019781.1

Gene Symbol
RPL6
Entrez Gene ID
6128
Gene Name
ribosomal protein L6
Gene Aliases
L6, SHUJUN-2, TAXREB107, TXREB1
UniGene
Hs.528668
Gene Chromosome Location
Chr.12: 112405181-112418850 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 NM_001320141.1 XM_017019781.1

SNP Detail

SNP ID Chromosome Location
rs987977948 Chr.12: 112418773-112418774 on Build GRCh38
rs966616210 Chr.12: 112418730-112418730 on Build GRCh38
rs540177251 Chr.12: 112418716-112418716 on Build GRCh38
rs538616585 Chr.12: 112418776-112418776 on Build GRCh38
rs377496292 Chr.12: 112418825-112418825 on Build GRCh38
rs554850776 Chr.12: 112418828-112418828 on Build GRCh38
rs543228277 Chr.12: 112418863-112418863 on Build GRCh38
rs58805176 Chr.12: 112418839-112418839 on Build GRCh38
rs375766559 Chr.12: 112418816-112418816 on Build GRCh38
rs897922842 Chr.12: 112418820-112418820 on Build GRCh38
rs1028386638 Chr.12: 112418799-112418799 on Build GRCh38
rs373537430 Chr.12: 112418795-112418795 on Build GRCh38
rs1032253135 Chr.12: 112418764-112418764 on Build GRCh38
rs940282387 Chr.12: 112418856-112418856 on Build GRCh38
rs954204477 Chr.12: 112418812-112418812 on Build GRCh38
rs542603894 Chr.12: 112418826-112418826 on Build GRCh38
rs942223575 Chr.12: 112418802-112418802 on Build GRCh38
rs913653431 Chr.12: 112418775-112418775 on Build GRCh38
rs1031665094 Chr.12: 112418787-112418787 on Build GRCh38
rs909449643 Chr.12: 112418801-112418801 on Build GRCh38
rs61941345 Chr.12: 112418789-112418789 on Build GRCh38
rs1030313327 Chr.12: 112418710-112418710 on Build GRCh38
rs957469684 Chr.12: 112418794-112418794 on Build GRCh38
rs953278097 Chr.12: 112418712-112418712 on Build GRCh38
rs997562896 Chr.12: 112418821-112418821 on Build GRCh38
rs999302793 Chr.12: 112418771-112418771 on Build GRCh38

View Full Product Details