Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 47234557-47234682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDB2
Entrez Gene ID
1643
Gene Name
damage specific DNA binding protein 2
Gene Aliases
DDBB, UV-DDB2, XPE
UniGene
Hs.700338
Gene Chromosome Location
Chr.11: 47214942-47239218 on Build GRCh38
Transcripts
NM_000107.2 NM_001300734.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4033132 c.689T>C p.M230T Chr.11: 47234659-47234659 on Build GRCh38
5529622 c.684G>A p.L228L Chr.11: 47234654-47234654 on Build GRCh38
7250703 c.606C>G p.I202M Chr.11: 47234576-47234576 on Build GRCh38
5838639 c.695G>A p.G232D Chr.11: 47234665-47234665 on Build GRCh38
6641246 c.643A>C p.M215L Chr.11: 47234613-47234613 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs764805641 Chr.11: 47234634-47234634 on Build GRCh38
rs201525567 Chr.11: 47234642-47234642 on Build GRCh38
rs530983301 Chr.11: 47234633-47234633 on Build GRCh38
rs202083037 Chr.11: 47234644-47234644 on Build GRCh38
rs370654581 Chr.11: 47234611-47234611 on Build GRCh38
rs759622121 Chr.11: 47234588-47234588 on Build GRCh38
rs144989465 Chr.11: 47234610-47234610 on Build GRCh38
rs754261384 Chr.11: 47234622-47234622 on Build GRCh38
rs781117402 Chr.11: 47234661-47234661 on Build GRCh38
rs4647750 Chr.11: 47234614-47234614 on Build GRCh38
rs761285630 Chr.11: 47234612-47234612 on Build GRCh38
rs753070223 Chr.11: 47234589-47234589 on Build GRCh38
rs755006840 Chr.11: 47234660-47234660 on Build GRCh38
rs930059864 Chr.11: 47234662-47234662 on Build GRCh38
rs149583624 Chr.11: 47234663-47234663 on Build GRCh38
rs962829260 Chr.11: 47234637-47234637 on Build GRCh38
rs779659936 Chr.11: 47234658-47234658 on Build GRCh38
rs139325563 Chr.11: 47234659-47234659 on Build GRCh38
rs774675310 Chr.11: 47234664-47234664 on Build GRCh38

View Full Product Details