Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 41748472-41748634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHOX2B
Entrez Gene ID
8929
Gene Name
paired like homeobox 2b
Gene Aliases
NBLST2, NBPhox, PMX2B
UniGene
Hs.87202
Gene Chromosome Location
Chr.4: 41744082-41748970 on Build GRCh38
Transcripts
NM_003924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3603898 c.132T>A p.Y44* Chr.4: 41748479-41748479 on Build GRCh38
5884023 c.119G>A p.S40N Chr.4: 41748492-41748492 on Build GRCh38
6984847 c.86C>T p.A29V Chr.4: 41748525-41748525 on Build GRCh38
6920424 c.56C>A p.A19D Chr.4: 41748555-41748555 on Build GRCh38
4771064 c.41A>G p.Y14C Chr.4: 41748570-41748570 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775101470 Chr.4: 41748569-41748569 on Build GRCh38
rs374160638 Chr.4: 41748500-41748500 on Build GRCh38
rs587778605 Chr.4: 41748552-41748552 on Build GRCh38
rs769573717 Chr.4: 41748566-41748566 on Build GRCh38
rs910430385 Chr.4: 41748539-41748539 on Build GRCh38
rs748767112 Chr.4: 41748572-41748572 on Build GRCh38
rs766500239 Chr.4: 41748497-41748497 on Build GRCh38
rs968453180 Chr.4: 41748559-41748559 on Build GRCh38
rs200971068 Chr.4: 41748541-41748541 on Build GRCh38
rs577333579 Chr.4: 41748577-41748577 on Build GRCh38
rs779251110 Chr.4: 41748512-41748512 on Build GRCh38
rs773982008 Chr.4: 41748578-41748578 on Build GRCh38
rs755290304 Chr.4: 41748506-41748506 on Build GRCh38
rs745833730 Chr.4: 41748554-41748554 on Build GRCh38
rs780786684 Chr.4: 41748550-41748550 on Build GRCh38
rs746854983 Chr.4: 41748533-41748533 on Build GRCh38
rs200038327 Chr.4: 41748521-41748521 on Build GRCh38
rs1043632230 Chr.4: 41748514-41748514 on Build GRCh38

View Full Product Details