Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 12720416-12720647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP9Y
Entrez Gene ID
8287
Gene Name
ubiquitin specific peptidase 9, Y-linked
Gene Aliases
DFFRY, SPGFY2
UniGene
Hs.598540
Gene Chromosome Location
Chr.Y: 12701231-12860844 on Build GRCh38
Transcripts
NM_004654.3 XM_017030078.1

SNP Detail

SNP ID Chromosome Location
rs867501709 Chr.Y: 12720593-12720593 on Build GRCh38
rs989102808 Chr.Y: 12720620-12720620 on Build GRCh38
rs747952575 Chr.Y: 12720556-12720556 on Build GRCh38
rs944253703 Chr.Y: 12720597-12720597 on Build GRCh38
rs369739862 Chr.Y: 12720546-12720546 on Build GRCh38
rs954047791 Chr.Y: 12720448-12720448 on Build GRCh38
rs778772298 Chr.Y: 12720548-12720548 on Build GRCh38

View Full Product Details