Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23415338-23415609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MHRT
Entrez Gene ID
104564225
Gene Name
myosin heavy chain-associated RNA transcript
Gene Aliases
Myheart
UniGene
Gene Chromosome Location
Chr.14: 23415450-23417595 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NR_126491.1 NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4850286 c.5197G>A p.D1733N Chr.14: 23415467-23415467 on Build GRCh38
4551398 c.5280G>A p.T1760T Chr.14: 23415384-23415384 on Build GRCh38
954740 c.5181G>T p.K1727N Chr.14: 23415483-23415483 on Build GRCh38
4551228 c.5230G>A p.A1744T Chr.14: 23415434-23415434 on Build GRCh38
4551240 c.5239G>A p.E1747K Chr.14: 23415425-23415425 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531250955 Chr.14: 23415442-23415442 on Build GRCh38
rs771626451 Chr.14: 23415361-23415361 on Build GRCh38
rs779595378 Chr.14: 23415444-23415444 on Build GRCh38
rs759357786 Chr.14: 23415429-23415429 on Build GRCh38
rs369453980 Chr.14: 23415521-23415521 on Build GRCh38
rs200303340 Chr.14: 23415421-23415421 on Build GRCh38
rs772148835 Chr.14: 23415374-23415374 on Build GRCh38
rs759194141 Chr.14: 23415540-23415540 on Build GRCh38
rs747252861 Chr.14: 23415486-23415486 on Build GRCh38
rs765784880 Chr.14: 23415585-23415585 on Build GRCh38
rs752456077 Chr.14: 23415526-23415527 on Build GRCh38
rs730880916 Chr.14: 23415410-23415410 on Build GRCh38
rs760355636 Chr.14: 23415547-23415547 on Build GRCh38
rs727505294 Chr.14: 23415385-23415385 on Build GRCh38
rs777325314 Chr.14: 23415434-23415434 on Build GRCh38
rs756851115 Chr.14: 23415584-23415584 on Build GRCh38
rs775803553 Chr.14: 23415380-23415380 on Build GRCh38
rs750973410 Chr.14: 23415586-23415586 on Build GRCh38
rs772504062 Chr.14: 23415463-23415463 on Build GRCh38
rs752406339 Chr.14: 23415433-23415433 on Build GRCh38
rs780076678 Chr.14: 23415477-23415477 on Build GRCh38
rs587779395 Chr.14: 23415475-23415476 on Build GRCh38
rs893202710 Chr.14: 23415412-23415412 on Build GRCh38
rs767027931 Chr.14: 23415430-23415430 on Build GRCh38
rs761917114 Chr.14: 23415512-23415512 on Build GRCh38
rs886050416 Chr.14: 23415456-23415456 on Build GRCh38
rs1001644180 Chr.14: 23415566-23415566 on Build GRCh38
rs770811309 Chr.14: 23415515-23415515 on Build GRCh38
rs867004523 Chr.14: 23415475-23415475 on Build GRCh38
rs878853839 Chr.14: 23415392-23415392 on Build GRCh38
rs876657885 Chr.14: 23415473-23415473 on Build GRCh38
rs548862977 Chr.14: 23415551-23415551 on Build GRCh38
rs397516240 Chr.14: 23415472-23415472 on Build GRCh38
rs910988779 Chr.14: 23415562-23415562 on Build GRCh38
rs962418187 Chr.14: 23415570-23415570 on Build GRCh38
rs374803185 Chr.14: 23415384-23415384 on Build GRCh38
rs753359198 Chr.14: 23415554-23415554 on Build GRCh38
rs148739072 Chr.14: 23415501-23415501 on Build GRCh38
rs730880814 Chr.14: 23415401-23415401 on Build GRCh38
rs997777221 Chr.14: 23415537-23415537 on Build GRCh38
rs776666954 Chr.14: 23415504-23415504 on Build GRCh38
rs776282574 Chr.14: 23415411-23415411 on Build GRCh38
rs545585809 Chr.14: 23415407-23415407 on Build GRCh38
rs144066768 Chr.14: 23415461-23415461 on Build GRCh38
rs149509691 Chr.14: 23415435-23415435 on Build GRCh38
rs1020453843 Chr.14: 23415518-23415518 on Build GRCh38
rs45609633 Chr.14: 23415541-23415541 on Build GRCh38
rs1011777797 Chr.14: 23415516-23415516 on Build GRCh38
rs367543052 Chr.14: 23415476-23415478 on Build GRCh38
rs730880813 Chr.14: 23415409-23415409 on Build GRCh38
rs775212799 Chr.14: 23415544-23415544 on Build GRCh38
rs1052787042 Chr.14: 23415366-23415366 on Build GRCh38
rs886044910 Chr.14: 23415491-23415493 on Build GRCh38
rs968070325 Chr.14: 23415536-23415536 on Build GRCh38
rs730880812 Chr.14: 23415458-23415458 on Build GRCh38
rs746278750 Chr.14: 23415363-23415363 on Build GRCh38
rs562465062 Chr.14: 23415419-23415419 on Build GRCh38
rs45582836 Chr.14: 23415362-23415362 on Build GRCh38

View Full Product Details