Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43600593-43600850 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRC
Entrez Gene ID
161497
Gene Name
stereocilin
Gene Aliases
DFNB16
UniGene
Hs.657395
Gene Chromosome Location
Chr.15: 43599243-43619669 on Build GRCh38
Transcripts
NM_153700.2 XM_011521278.2 XM_017021954.1 XM_017021955.1 XM_017021956.1 XM_017021957.1 XM_017021958.1 XM_017021959.1 XM_017021962.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5643524 c.4849G>A p.A1617T Chr.15: 43600678-43600678 on Build GRCh38
3816155 c.4857C>G p.L1619L Chr.15: 43600670-43600670 on Build GRCh38
6792394 c.4851A>G p.A1617A Chr.15: 43600676-43600676 on Build GRCh38
6457556 c.4886C>G p.S1629C Chr.15: 43600641-43600641 on Build GRCh38
1373058 c.4903G>T p.V1635F Chr.15: 43600624-43600624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1010941642 Chr.15: 43600812-43600812 on Build GRCh38
rs763385671 Chr.15: 43600830-43600830 on Build GRCh38
rs775312704 Chr.15: 43600716-43600716 on Build GRCh38
rs369417288 Chr.15: 43600685-43600685 on Build GRCh38
rs776707430 Chr.15: 43600622-43600622 on Build GRCh38
rs761361123 Chr.15: 43600659-43600659 on Build GRCh38
rs375284306 Chr.15: 43600639-43600639 on Build GRCh38
rs12438025 Chr.15: 43600649-43600649 on Build GRCh38
rs768328048 Chr.15: 43600722-43600722 on Build GRCh38
rs1052725790 Chr.15: 43600790-43600790 on Build GRCh38
rs560371288 Chr.15: 43600797-43600797 on Build GRCh38
rs374759204 Chr.15: 43600733-43600733 on Build GRCh38
rs893794893 Chr.15: 43600803-43600803 on Build GRCh38
rs930367421 Chr.15: 43600694-43600694 on Build GRCh38
rs764784468 Chr.15: 43600734-43600734 on Build GRCh38
rs867795318 Chr.15: 43600778-43600778 on Build GRCh38
rs770037300 Chr.15: 43600704-43600704 on Build GRCh38
rs574386839 Chr.15: 43600793-43600793 on Build GRCh38
rs772061973 Chr.15: 43600640-43600640 on Build GRCh38
rs760405894 Chr.15: 43600717-43600717 on Build GRCh38
rs184909019 Chr.15: 43600682-43600682 on Build GRCh38
rs2447206 Chr.15: 43600810-43600810 on Build GRCh38
rs774483463 Chr.15: 43600633-43600633 on Build GRCh38
rs139683375 Chr.15: 43600664-43600664 on Build GRCh38
rs775159378 Chr.15: 43600641-43600641 on Build GRCh38
rs376948482 Chr.15: 43600712-43600712 on Build GRCh38
rs961928299 Chr.15: 43600632-43600632 on Build GRCh38
rs776304785 Chr.15: 43600724-43600724 on Build GRCh38
rs12437957 Chr.15: 43600695-43600695 on Build GRCh38
rs758779420 Chr.15: 43600773-43600773 on Build GRCh38
rs770943687 Chr.15: 43600628-43600628 on Build GRCh38
rs778993261 Chr.15: 43600705-43600705 on Build GRCh38
rs765702125 Chr.15: 43600661-43600661 on Build GRCh38
rs753336088 Chr.15: 43600655-43600655 on Build GRCh38
rs745860856 Chr.15: 43600707-43600707 on Build GRCh38
rs540126867 Chr.15: 43600795-43600795 on Build GRCh38
rs2915791 Chr.15: 43600624-43600624 on Build GRCh38
rs760304841 Chr.15: 43600645-43600645 on Build GRCh38
rs927800686 Chr.15: 43600617-43600617 on Build GRCh38
rs999717999 Chr.15: 43600786-43600786 on Build GRCh38
rs771905551 Chr.15: 43600711-43600711 on Build GRCh38
rs750994204 Chr.15: 43600663-43600663 on Build GRCh38
rs367587412 Chr.15: 43600741-43600741 on Build GRCh38
rs189488577 Chr.15: 43600798-43600798 on Build GRCh38
rs751876108 Chr.15: 43600670-43600670 on Build GRCh38
rs751903876 Chr.15: 43600831-43600831 on Build GRCh38
rs1041170555 Chr.15: 43600770-43600770 on Build GRCh38
rs768775965 Chr.15: 43600615-43600615 on Build GRCh38
rs2447207 Chr.15: 43600821-43600821 on Build GRCh38

View Full Product Details