Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 12721995-12722268 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP9Y
Entrez Gene ID
8287
Gene Name
ubiquitin specific peptidase 9, Y-linked
Gene Aliases
DFFRY, SPGFY2
UniGene
Hs.598540
Gene Chromosome Location
Chr.Y: 12701231-12860844 on Build GRCh38
Transcripts
NM_004654.3 XM_017030078.1

SNP Detail

SNP ID Chromosome Location
rs542362399 Chr.Y: 12722113-12722113 on Build GRCh38
rs781217987 Chr.Y: 12722228-12722228 on Build GRCh38
rs954097233 Chr.Y: 12722217-12722217 on Build GRCh38
rs769114689 Chr.Y: 12722153-12722153 on Build GRCh38
rs755711770 Chr.Y: 12722180-12722180 on Build GRCh38
rs749384578 Chr.Y: 12722230-12722239 on Build GRCh38
rs779626695 Chr.Y: 12722196-12722196 on Build GRCh38
rs747355658 Chr.Y: 12722127-12722127 on Build GRCh38
rs767193864 Chr.Y: 12722148-12722148 on Build GRCh38
rs753218919 Chr.Y: 12722213-12722213 on Build GRCh38

View Full Product Details