Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23413696-23413951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH7
Entrez Gene ID
4625
Gene Name
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases
CMD1S, CMH1, MPD1, MYHCB, SPMD, SPMM
UniGene
Hs.719946
Gene Chromosome Location
Chr.14: 23412738-23435686 on Build GRCh38
Transcripts
NM_000257.3 XM_017021340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4450368 c.5656-1G>A p.? Chr.14: 23413894-23413894 on Build GRCh38
5694643 c.5726G>A p.R1909Q Chr.14: 23413823-23413823 on Build GRCh38
2031714 c.5737G>A p.A1913T Chr.14: 23413812-23413812 on Build GRCh38
2031715 c.5730G>A p.A1910A Chr.14: 23413819-23413819 on Build GRCh38
3495199 c.5715G>A p.E1905E Chr.14: 23413834-23413834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897499056 Chr.14: 23413727-23413727 on Build GRCh38
rs747451109 Chr.14: 23413812-23413812 on Build GRCh38
rs397516252 Chr.14: 23413832-23413832 on Build GRCh38
rs755706526 Chr.14: 23413776-23413776 on Build GRCh38
rs752553589 Chr.14: 23413775-23413775 on Build GRCh38
rs199651194 Chr.14: 23413903-23413903 on Build GRCh38
rs754705892 Chr.14: 23413766-23413766 on Build GRCh38
rs781212482 Chr.14: 23413902-23413902 on Build GRCh38
rs199975761 Chr.14: 23413744-23413744 on Build GRCh38
rs540350007 Chr.14: 23413885-23413885 on Build GRCh38
rs778562229 Chr.14: 23413862-23413862 on Build GRCh38
rs397516254 Chr.14: 23413809-23413809 on Build GRCh38
rs988052048 Chr.14: 23413840-23413840 on Build GRCh38
rs776550740 Chr.14: 23413755-23413755 on Build GRCh38
rs763606475 Chr.14: 23413850-23413850 on Build GRCh38
rs45523233 Chr.14: 23413831-23413831 on Build GRCh38
rs45570635 Chr.14: 23413846-23413846 on Build GRCh38
rs760334246 Chr.14: 23413927-23413927 on Build GRCh38
rs121913649 Chr.14: 23413847-23413847 on Build GRCh38
rs730880918 Chr.14: 23413763-23413763 on Build GRCh38
rs1014585705 Chr.14: 23413726-23413726 on Build GRCh38
rs397516256 Chr.14: 23413787-23413787 on Build GRCh38
rs786204385 Chr.14: 23413778-23413778 on Build GRCh38
rs763957408 Chr.14: 23413930-23413930 on Build GRCh38
rs775031326 Chr.14: 23413722-23413722 on Build GRCh38
rs373150382 Chr.14: 23413720-23413720 on Build GRCh38
rs138110910 Chr.14: 23413795-23413795 on Build GRCh38
rs763067471 Chr.14: 23413918-23413918 on Build GRCh38
rs775523172 Chr.14: 23413925-23413925 on Build GRCh38
rs761750159 Chr.14: 23413822-23413822 on Build GRCh38
rs770065133 Chr.14: 23413911-23413911 on Build GRCh38
rs397516250 Chr.14: 23413897-23413897 on Build GRCh38
rs397516253 Chr.14: 23413823-23413823 on Build GRCh38
rs187073962 Chr.14: 23413845-23413845 on Build GRCh38
rs376965306 Chr.14: 23413913-23413913 on Build GRCh38
rs539290591 Chr.14: 23413788-23413788 on Build GRCh38
rs756655803 Chr.14: 23413860-23413860 on Build GRCh38
rs776940948 Chr.14: 23413819-23413819 on Build GRCh38
rs375404554 Chr.14: 23413889-23413889 on Build GRCh38
rs727503240 Chr.14: 23413859-23413859 on Build GRCh38
rs869025483 Chr.14: 23413805-23413808 on Build GRCh38
rs397516255 Chr.14: 23413800-23413800 on Build GRCh38
rs397516251 Chr.14: 23413901-23413901 on Build GRCh38
rs200231434 Chr.14: 23413898-23413898 on Build GRCh38
rs767300277 Chr.14: 23413770-23413770 on Build GRCh38
rs397516257 Chr.14: 23413762-23413762 on Build GRCh38
rs577825712 Chr.14: 23413922-23413922 on Build GRCh38
rs1015495430 Chr.14: 23413895-23413895 on Build GRCh38
rs146796870 Chr.14: 23413824-23413824 on Build GRCh38
rs202232153 Chr.14: 23413758-23413758 on Build GRCh38
rs886042307 Chr.14: 23413901-23413902 on Build GRCh38
rs868275632 Chr.14: 23413865-23413865 on Build GRCh38
rs200921000 Chr.14: 23413810-23413810 on Build GRCh38
rs546582990 Chr.14: 23413749-23413749 on Build GRCh38
rs556403696 Chr.14: 23413921-23413921 on Build GRCh38
rs370553746 Chr.14: 23413745-23413745 on Build GRCh38
rs764434831 Chr.14: 23413752-23413752 on Build GRCh38
rs200728597 Chr.14: 23413813-23413813 on Build GRCh38
rs560070845 Chr.14: 23413894-23413894 on Build GRCh38
rs730880727 Chr.14: 23413888-23413888 on Build GRCh38
rs727504384 Chr.14: 23413853-23413853 on Build GRCh38
rs730880892 Chr.14: 23413890-23413890 on Build GRCh38
rs777524474 Chr.14: 23413784-23413784 on Build GRCh38

View Full Product Details