Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112472863-112473118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN11
Entrez Gene ID
5781
Gene Name
protein tyrosine phosphatase, non-receptor type 11
Gene Aliases
BPTP3, CFC, JMML, METCDS, NS1, PTP-1D, PTP2C, SH-PTP2, SH-PTP3, SHP2
UniGene
Hs.506852
Gene Chromosome Location
Chr.12: 112418732-112509913 on Build GRCh38
Transcripts
NM_002834.3 NM_080601.1 XM_006719526.2 XM_011538613.2 XM_017019722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5635575 c.811C>G p.Q271E Chr.12: 112472998-112472998 on Build GRCh38
6758805 c.786C>G p.L262L Chr.12: 112472973-112472973 on Build GRCh38
1948751 c.794G>A p.R265Q Chr.12: 112472981-112472981 on Build GRCh38
5945279 c.853T>C p.F285L Chr.12: 112473040-112473040 on Build GRCh38
6919376 c.844A>G p.I282V Chr.12: 112473031-112473031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763617831 Chr.12: 112472975-112472975 on Build GRCh38
rs200179109 Chr.12: 112472918-112472918 on Build GRCh38
rs751823249 Chr.12: 112472907-112472907 on Build GRCh38
rs121918456 Chr.12: 112473023-112473023 on Build GRCh38
rs397507531 Chr.12: 112473040-112473040 on Build GRCh38
rs749236518 Chr.12: 112472921-112472921 on Build GRCh38
rs766531368 Chr.12: 112472980-112472980 on Build GRCh38
rs547083802 Chr.12: 112473097-112473097 on Build GRCh38
rs397507530 Chr.12: 112473033-112473033 on Build GRCh38
rs886048966 Chr.12: 112473048-112473048 on Build GRCh38
rs397507527 Chr.12: 112472989-112472989 on Build GRCh38
rs987889795 Chr.12: 112473081-112473081 on Build GRCh38
rs755104848 Chr.12: 112472988-112472988 on Build GRCh38
rs397507525 Chr.12: 112472968-112472968 on Build GRCh38
rs376607329 Chr.12: 112472981-112472981 on Build GRCh38
rs755288216 Chr.12: 112472910-112472910 on Build GRCh38
rs745489506 Chr.12: 112472949-112472949 on Build GRCh38
rs397507523 Chr.12: 112472954-112472954 on Build GRCh38
rs781016088 Chr.12: 112472913-112472913 on Build GRCh38
rs397507529 Chr.12: 112473031-112473031 on Build GRCh38
rs770950143 Chr.12: 112473092-112473092 on Build GRCh38
rs571488248 Chr.12: 112473039-112473039 on Build GRCh38
rs1032100840 Chr.12: 112473059-112473059 on Build GRCh38
rs773417679 Chr.12: 112472966-112472966 on Build GRCh38
rs752966396 Chr.12: 112473001-112473001 on Build GRCh38
rs372220853 Chr.12: 112473052-112473052 on Build GRCh38
rs886041495 Chr.12: 112473032-112473032 on Build GRCh38
rs751001606 Chr.12: 112472894-112472894 on Build GRCh38
rs374866811 Chr.12: 112473071-112473071 on Build GRCh38
rs397507526 Chr.12: 112472972-112472972 on Build GRCh38
rs777603059 Chr.12: 112473014-112473014 on Build GRCh38
rs775232682 Chr.12: 112472955-112472955 on Build GRCh38
rs765642157 Chr.12: 112472969-112472969 on Build GRCh38
rs200165689 Chr.12: 112472919-112472919 on Build GRCh38
rs917536914 Chr.12: 112472984-112472984 on Build GRCh38
rs771646462 Chr.12: 112473050-112473050 on Build GRCh38
rs1007360259 Chr.12: 112473058-112473058 on Build GRCh38
rs397507524 Chr.12: 112472957-112472958 on Build GRCh38
rs771672596 Chr.12: 112472950-112472950 on Build GRCh38
rs794727858 Chr.12: 112472993-112472993 on Build GRCh38
rs749220083 Chr.12: 112473019-112473019 on Build GRCh38
rs933129070 Chr.12: 112473000-112473000 on Build GRCh38
rs747847358 Chr.12: 112472914-112472914 on Build GRCh38
rs746692191 Chr.12: 112473057-112473057 on Build GRCh38
rs758490437 Chr.12: 112472903-112472903 on Build GRCh38
rs876657567 Chr.12: 112473018-112473018 on Build GRCh38
rs397516809 Chr.12: 112472961-112472961 on Build GRCh38
rs757228212 Chr.12: 112473021-112473021 on Build GRCh38
rs532605873 Chr.12: 112473084-112473084 on Build GRCh38
rs778374689 Chr.12: 112473034-112473034 on Build GRCh38
rs550231049 Chr.12: 112472923-112472923 on Build GRCh38
rs397507528 Chr.12: 112473011-112473011 on Build GRCh38
rs561136680 Chr.12: 112472906-112472906 on Build GRCh38
rs773918720 Chr.12: 112472944-112472944 on Build GRCh38

View Full Product Details