Gene Transcripts

Gene Symbol
Prmt10
Entrez Gene ID
102182
Gene Name
protein arginine methyltransferase 10 (putative)
Gene Aliases
AI931714, Prmt9
Chromosome Location
Chr. 8: 77549387 - 77581338 on Build GRCm38
UniGene ID
Mm.197630
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081240.3 NP_001074709.1 9 1726
GenBank mRNA AK045315.1 - 9 1716
AK154546.1 - 9 1670
AK161023.1 - 9 1635

View Full Product Details