Gene Transcripts

Gene Symbol
Rcbtb2
Entrez Gene ID
105670
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2
Gene Aliases
2610028E02Rik, 2810420M18Rik, AI448345, AW240694, Chc1l, RC/BTB2
Chromosome Location
Chr. 14: 73138773 - 73184055 on Build GRCm38
UniGene ID
Mm.280068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170694.1 NP_001164165.1 15 1818
NM_134083.4 NP_598844.3 15 1813
GenBank mRNA AK031242.1 - 15 1779
AK031248.1 - 15 1784
AK147784.1 - 16 1819
AK160212.1 - 3 1655
AK160881.1 - 16 2005
AK183696.1 - 4 379
AK191301.1 - 4 370
AK210358.1 - 4 317
AK217036.1 - 4 317
BC003224.1 - 13 1611 3482729
JN086338.1 - 14 1734
JN086339.1 - 15 1972

View Full Product Details