Gene Transcripts

Gene Symbol
Fntb
Entrez Gene ID
110606
Gene Name
farnesyltransferase, CAAX box, beta
Gene Aliases
2010013E13Rik, AA409500
Chromosome Location
Chr. 12: 76837398 - 76921412 on Build GRCm38
UniGene ID
Mm.151174
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145927.2 NP_666039.1 7 692
GenBank mRNA AK089911.1 - 7 722
BC031417.1 - 7 692 3154478

View Full Product Details