Gene Transcripts

Gene Symbol
Brdt
Entrez Gene ID
114642
Gene Name
bromodomain, testis-specific
Gene Aliases
7420412D09Rik, Brd6, Fsrg3
Chromosome Location
Chr. 5: 107331091 - 107387061 on Build GRCm38
UniGene ID
Mm.182836
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079873.1 NP_001073342.1 7 1110
NM_054054.2 NP_473395.2 7 1110
GenBank mRNA AB208640.1 - 5 803
AF358660.1 - 7 1145
AK077137.1 - 7 1130
BY726053.1 - 3 436

View Full Product Details