Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007468.2 | NP_031494.2 | 3 | 736 | |
GenBank mRNA | AK004856.1 | - | 3 | 519 | |
AK161535.1 | - | 3 | 519 | ||
AK168687.1 | - | 3 | 520 | ||
BC010769.1 | - | 3 | 529 | 4240796 | |
BC050149.1 | - | 3 | 494 | 30013747 | |
M64248.1 | - | 3 | 518 | ||
M64249.1 | - | 3 | 518 | ||
M64250.1 | - | 3 | 518 |