Gene Transcripts
- Gene Symbol
- Arhgap31
- Entrez Gene ID
- 12549
- Gene Name
- Rho GTPase activating protein 31
- Gene Aliases
- 5830477L08Rik, AU041750, Cdgap, D330026I07Rik
- Chromosome Location
- Chr. 16: 38598343 - 38713035 on Build GRCm38
- UniGene ID
- Mm.370848
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020260.2 |
NP_064656.2
|
2, 3 |
570 |
|
GenBank mRNA |
AF151363.1 |
- |
2, 3 |
211 |
|
AK018057.1 |
- |
2, 3 |
436 |
|
AK138417.1 |
- |
2, 3 |
810 |
|
AK149975.1 |
- |
2, 3 |
406 |
|
AK150109.1 |
- |
2, 3 |
406 |
|
AK155729.1 |
- |
2, 3 |
390 |
|
AK170685.1 |
- |
2, 3 |
390 |
|
BB865916.1 |
- |
2, 3 |
572 |
|
BC138870.1 |
- |
2, 3 |
375 |
8861892 |
View Full Product Details