Gene Transcripts

Gene Symbol
Hsd17b4
Entrez Gene ID
15488
Gene Name
hydroxysteroid (17-beta) dehydrogenase 4
Gene Aliases
17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
Chromosome Location
Chr. 18: 50128201 - 50196270 on Build GRCm38
UniGene ID
Mm.277857
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008292.4 NP_032318.2 14, 15 1403
GenBank mRNA AK004866.1 - 14, 15 1403
AK088381.1 - 14, 15 1378
AK147182.1 - 14, 15 1375
AK152532.1 - 14, 15 1298
AK161655.1 - 14, 15 1298
AK166801.1 - 14, 15 1316
AK167060.1 - 14, 15 1353
AK169077.1 - 14, 15 1305
AK187517.1 - 2, 3 294
AK216937.1 - 2, 3 294
BC022175.1 - 14, 15 1276 4209394

View Full Product Details