Gene Transcripts
- Gene Symbol
- Apcs
- Entrez Gene ID
- 20219
- Gene Name
- serum amyloid P-component
- Gene Aliases
- Sap
- Chromosome Location
- Chr. 1: 172893961 - 172895054 on Build GRCm38
- UniGene ID
- Mm.330510
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
Y00426.1 |
- |
2 |
622 |
|
View Full Product Details