Gene Transcripts

Gene Symbol
Aftph
Entrez Gene ID
216549
Gene Name
aftiphilin
Gene Aliases
Chromosome Location
Chr. 11: 20685084 - 20742841 on Build GRCm38
UniGene ID
Mm.356580
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252503.2 NP_001239432.1 2 2048
NM_001290545.1 NP_001277474.1 2 2048
NM_181411.4 NP_852076.1 2 2048
GenBank mRNA AK018647.1 - 1 187
AK029681.1 - 2 1947
AK030572.1 - 2 1992
AK047225.1 - 1 1112
AK136815.1 - 1 1300
AK138793.1 - 2 1970
BC004630.1 - 1 337
BC052036.1 - 2 1976 5702534

View Full Product Details