Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011639.3 | NP_035769.1 | 5 | 968 | |
GenBank mRNA | AF097511.1 | - | 5 | 861 | |
AF116823.1 | - | 5 | 963 | ||
AK010435.1 | - | 5 | 969 | ||
AK146823.1 | - | 5 | 918 | ||
AK159387.1 | - | 5 | 937 | ||
AK166024.1 | - | 5 | 1132 | ||
BC138685.1 | - | 5 | 900 | 8861706 | |
BC138686.1 | - | 5 | 900 | 8861707 |