Gene Transcripts

Gene Symbol
Hrh4
Entrez Gene ID
225192
Gene Name
histamine receptor H4
Gene Aliases
AXOR35, BG26, GPCR105, GPRv53, H4, H4R, HH4R
Chromosome Location
Chr. 18: 13006982 - 13023701 on Build GRCm38
UniGene ID
Mm.207073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153087.2 NP_694727.1 3 782
GenBank mRNA AF358859.1 - 3 782
BC111862.1 - 2 617 40046148

View Full Product Details