Gene Transcripts

Gene Symbol
Coq4
Entrez Gene ID
227683
Gene Name
coenzyme Q4 homolog (yeast)
Gene Aliases
A330041N06, D2Ertd97e
Chromosome Location
Chr. 2: 29788226 - 29797743 on Build GRCm38
UniGene ID
Mm.245948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178693.4 NP_848808.1 5, 6 559
GenBank mRNA AK039422.1 - 5, 6 560
AK039676.1 - 5, 6 560
AK043574.1 - 5, 6 535
AK050459.1 - 5, 6 569
BC139373.1 - 5, 6 602 8862395
BC139376.1 - 5, 6 599 8862398
BC145615.1 - 5, 6 573

View Full Product Details